Thermo Scientific™

POROS™ CaptureSelect™ AAV9 Affinity Resin

Catalog number: A27357
Have Questions?
Thermo Scientific™

POROS™ CaptureSelect™ AAV9 Affinity Resin

Catalog number: A27357
Have Questions?
Catalog Number
A27357
Unit Size
10,000 mL
Price (USD)
Availability
-
Quantity
-
Catalog Number
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
250 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,000 mL
Price (USD)
Availability
-
Quantity
-
Catalog Number
Unit Size
5,000 mL
Price (USD)
Availability
-
Quantity
-
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A2735710,000 mL
--
A2735410 mL
-
A2735325 mL
-
A2735650 mL
-
A27355250 mL
-
A273591,000 mL
--
A273585,000 mL
--
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

POROS™ CaptureSelect™ AAV9 high performance affinity resin is engineered to address the high selectivity and capacity requirements for the purification of viral vectors used for gene therapy applications and is specifically intended for the purification of adeno-associated virus type 9 (AAV9).

Features of POROS CaptureSelect AAV9 resin include:

  • One-step AAV9 purification from crude material with high purity and yield
  • High specificity and capacity, which helps reduce the process volume significantly for subsequent steps and maximizes yield
  • Low operating back pressure and linear pressure versus flow responses driving flexible scalability
  • Superior capacity maintained at high flow rates
  • Rigid polymeric bead with covalent surface chemistry for easier handling and packing, with superior physical and chemical stability enabling a more robust downstream purification process
  • Non-animal derived—the ligand is a 13-kDa single-domain fragment that comprises the 3 CDRs that form the antigen-binding domain and is efficiently produced by the yeast Saccharomyces cerevisiae in a production process free of animal components.

POROS CaptureSelect AAV9 affinity resin specifically binds AAV9 and enables a robust, fast, and efficient purification process with excellent purity and yield obtained in one step. The use of this affinity resin will enable significant improvement to downstream processing by reducing the purification steps and maximizing productivity, for scalability and processing consistency.

Main characteristics:

  • Matrix: cross-linked poly(styrene-divinylbenzene)
  • Average particle size: 50 μm
  • Ligand: CaptureSelect AAV9 affinity ligand
  • Ligand coupling method: carbonyldiimidazole (CDI)
  • Binding capacity: >1x1014 genome copies/mL of resin
  • Elution conditions: acidic elution with 0.1 M glycine, pH 3.0
  • Mechanical resistance: 100 bar (1,450 psi; 10 MPa)
  • Formulation buffer: 18% (v/v) ethanol

Specifications

Binding Capacity
>1 x 1014 genome copies/mL of resin
Matrix
Cross-linked poly(styrene-divinylbenzene)
Shipping Condition
Wet Ice
Particle Size
50 μm
Form
Liquid Suspension
Product Line
CaptureSelect™, POROS™
Quantity
10 L
Type
Resin
Ligand Type
AAV9 affinity ligand

Contents & Storage

Store at 2°C to 8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text