ProteinSEQ™ HCP Core Kit
To receive future technical notifications regarding this product, please click HERE to register
Product Image
Applied Biosystems™
ProteinSEQ™ HCP Core Kit
The ProteinSEQ™ HCP Core Kit is a customizable kit that enables users to customize their ProteinSEQ host cell protein measurementsRead more
Have Questions?
Catalog NumberQuantity
A27600200 Reactions
Catalog number A27600
Price (USD)
-
Request A Quote
Quantity:
200 Reactions
The ProteinSEQ™ HCP Core Kit is a customizable kit that enables users to customize their ProteinSEQ host cell protein measurements for their own cell lines. It contains buffers and reagents needed to perform 200 PCR reactions to quantify host cell proteins of any cell line for which there is an antibody available.
Features include:
• Precise—quantitate host cell protein contaminants with excellent precision
• Fast—helps accelerate process development with more valid test results per run
• Efficient—helps reduce hands-on time with less sample dilution and preparation
• Reproducible—achieve unprecedented productivity with 4-log dynamic range
ProteinSEQ CHO HCP quantitation
The ProteinSEQ assay platform uses qPCR technology to measure protein process contaminants with extraordinary sensitivity and unparalleled dynamic range. Host cell protein contaminants from any cell line can be measured easily and effectively by incorporating any anti-HCP antibody into the ProteinSEQ workflow.
Streamlined workflow
The ProteinSEQ HCP Core Kit is the only instrument platform-based HCP quantitation solution to deliver a customizable HCP assay with class-leading sensitivity and 4-log dynamic range. The kit helps accelerate laboratory workflows because the number of sample dilutions and replicates is reduced in each run.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayProtein Quantitation Assay
Detection MethodPrimer-Probe Detection
For Use With (Equipment)7500 Fast System
Shipping ConditionApproved for shipment on Wet or Dry Ice
Product LineProteinSEQ™
Quantity200 Reactions
For Use With (Application)Proximity Ligation Assay
TypeHCP Core Kit
FormatKit
Unit Size200 reactions
Contents & Storage
100 mL ProteinSEQ Wash Buffer, store at 2–8°C
50 mL CHO HCO ProteinSEQ Diluent, store at 2–8°C
7 mL ProteinSEQ Elution Buffer, store at 2–8°C
6 mL Fast Master Mix (2X), store at 2–8°C
600 μL ProteinSEQ Ligation and Assay Mix, store at -5 to -30°C
55 μL ProteinSEQ Ligase, store at -5 to -30°C
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link