Ion 520™ Chip Kit
Ion 520™ Chip Kit
Ion Torrent™

Ion 520™ Chip Kit

Green features
The Ion 520 Chip Kit contains 8 barcoded chips for sample tracking and sequencing with the Ion S5 and IonRead more
Have Questions?
Catalog NumberQuantity
A277628 chips
Catalog number A27762
Price (USD)
4,850.00
Each
In stock
Add to cart
Quantity:
8 chips
Price (USD)
4,850.00
Each
Add to cart
The Ion 520 Chip Kit contains 8 barcoded chips for sample tracking and sequencing with the Ion S5 and Ion S5 XL Sequencing Systems. The Ion 520 Chip electronically detects polymerase-driven base incorporation without the use of fluorescence. By eliminating the use of an optical detection system, this advance in next-generation sequencing technology allows for rapid sequencing times in as little as 2.5 hours for 200 bp sequencing.

The Ion 520 Chip Kit offers:

• Sample tracking functionality from an integrated barcode
• A rapid run time of only 2.5–4 hours, generating 3–6 million reads, depending on application
• Read lengths up to 600 bp to support a variety of research applications, including targeted gene sequencing and microbial sequencing
• Easy handling and loading of templated products for sequencing
• Compatibility with current library preparation methods

Notes:
This kit is only compatible with the Ion S5 and Ion S5 XL systems.
Use this kit with the Ion 510 & Ion 520 & Ion 530 Kit-Chef, the Ion 520 & Ion 530 ExT Kit-Chef, and the Ion 520 & Ion 530 Kit-OT2.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Ion GeneStudio S5 System, Ion S5™ XL System
Green FeaturesSustainable disposal
Number of Reads per Chip3-6M
Product TypeChip Kit
Quantity8 chips
Shipping ConditionRoom Temperature
Product LineIon 520™
Unit SizeEach
Contents & Storage
Store at room temperature

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection