Ion Torrent™

Ion 540™ Chip Kit

Catalog number: A27765
Have Questions?
Ion Torrent™

Ion 540™ Chip Kit

Catalog number: A27765
Have Questions?
Catalog Number
A27765
Unit Size
4 chips
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
8 chips
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A277654 chips
-
A277668 chips
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ion 540™ Chip Kit contains 8 barcoded chips for sample tracking and sequencing with the Ion S5™ and Ion S5™ XL Sequencing Systems. The Ion 540 Chip electronically detects polymerase-driven base incorporation without the use of fluorescence. By eliminating the use of an optical detection system, this advance in next-generation sequencing technology allows for rapid sequencing times of as little as 2.5 hours for 200 bp sequencing.

The Ion 540 Chip Kit offers:

• Sample tracking functionality from an integrated barcode
• A rapid run time of as little as 2.5 hours for 200 bp sequencing, generating 60–80 million reads
• Support for a variety of research applications, including sequencing of up to three human exomes, transcriptome profiling, and targeted resequencing of gene panels
• Easy handling and loading of templated products for sequencing
• Compatibility with current library preparation methods

Notes:
This kit is only compatible with the Ion S5 and Ion S5 XL systems.
Use this kit with the Ion 540 Kit-Chef and the Ion 540 Kit-OT2.

Specifications

Product Line
Ion 540™
Format
Kit
For Use With (Application)
Sequencing
Green Features
Sustainable disposal
Number of Reads per Chip
60-80M
Product Type
Chip Kit
Quantity
4 Chips
Sequencing Type
Genome and DNA Sequencing
Shipping Condition
Room Temperature
Species
Human

Contents & Storage

Store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text