Informational:cGMP-banked ExpiCHO-S cells now available. Click HERE for product information.
Gibco™

ExpiFectamine™ CHO Transfection Kit

Catalog number: A29130
Have Questions?
Gibco™

ExpiFectamine™ CHO Transfection Kit

Catalog number: A29130
Have Questions?
Catalog Number
A29130
Unit Size
For 10 L of culture
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
For 1 L of culture
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
For 50 L of culture
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A29130For 10 L of culture
-
A29129For 1 L of culture
-
A29131For 50 L of culture
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The ExpiFectamine™ CHO Transfection Kit is a core component of the ExpiCHO™ Expression System Kit (Cat. No. A29133). It is designed for transient transfection of high-density cultures of Chinese hamster ovary (CHO) cells. The high-efficiency, cationic transfection reagent, coupled with an optimized transfection enhancer and culture feed, will power the highest possible level of protein expression from ExpiCHO-S™ Cells (Cat. No. A29127) cultured in ExpiCHO™ Expression Medium (Cat. No. A2910001).

The ExpiFectamine CHO Transfection Kit:
• Achieves greater than 90% transfection efficiency of high-density suspension CHO cell culture, with matching transfection enhancer and culture feed to boost transfection performance and protein expression
• Requires one-third to one-half less plasmid DNA than other transfection reagents while achieving superior expression performance
• Provides options for three transient expression protocols for maximal work flexibility while still fitting into the same overall workflow used by other transient expression systems, such as FreeStyle™ MAX CHO and Expi293™
• Enables robust and reproducible transfection results, giving you greater confidence in your results
• Demonstrates linear scalability of transfections from culture volumes of <1 mL to >10 L

The ExpiFectamine CHO Transfection Kit is specifically designed to work with ExpiCHO-S Cells in the ExpiCHO™ Expression System Kit. The transfection kit supplies the cationic ExpiFectamine™ CHO Transfection Reagent, ExpiFectamine™ CHO Transfection Enhancer, and ExpiCHO Feed.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Classification
Animal Origin-Free, Chemically Defined
Description
Kit Includes: 32 mL ExpiFectamine™ CHO Reagent, 60 mL ExpiFectamine™ CHO Enhancer, 3.3 L ExpiCHO Feed
Cell Type
Ovarian Cells, Established Cell Lines
Transfection Technique
Lipid-based Transfection
Product Line
ExpiFectamine™
Product Type
CHO Transfection Kit
Quantity
For 10L of culture
Shipping Condition
Room Temperature

Contents & Storage

32 mL ExpiFectamine CHO Reagent
60 mL ExpiFectamine CHO Enhancer
3.3 L ExpiCHO Feed

Sufficient for transfection of 10 L of culture

Store in the dark at 2 -8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text