Applied Biosystems™

7500 Fast Precision Plate Holder, for 0.1 mL tube strips

Catalog number: A29252
Have Questions?
Applied Biosystems™

7500 Fast Precision Plate Holder, for 0.1 mL tube strips

Catalog number: A29252
Have Questions?
Catalog Number
A29252
Unit Size
1 each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A292521 each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems™ 7500 Fast Precision Plate Holder contains one precision plate holder to be used with 0.1 ml tube strips on the 7500 Fast System, and the recommended usage procedure. Note: A29252 replaces Cat. No. 4403809.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
7500 Fast System
Product Type
Precision Plate Holder
Shipping Condition
Room Temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text