Invitrogen™

Precision gRNA Synthesis Kit

Catalog number: A29377
Have Questions?
Invitrogen™

Precision gRNA Synthesis Kit

Catalog number: A29377
Have Questions?
Catalog Number
A29377
Unit Size
25 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A2937725 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Precision gRNA Synthesis Kit is a complete system for rapid synthesis of guide RNA (gRNA) ready to complex with TrueCut™ Cas9 Protein v2 for transfection-ready Cas9 protein/gRNA ribonucleoprotein (Cas9 RNP). This Cas9 RNP format, with our TrueCut Cas9 Protein v2, has been tested in a variety of suspension and adherent cell lines with >70% cleavage efficiencies and no indications of toxicity. Starting with two short single-stranded oligos that code for the target sequence, the gRNA template is assembled with a T7 promoter in a short ‘one-pot’ PCR reaction. The assembled product is then used as template in an in vitro transcription (IVT) reaction followed by a rapid purification step, yielding transfection-ready gRNA in as little as four hours. Resulting gRNA can also be co-transfected with our ready-to-transfect Invitrogen™ CRISPR Nuclease mRNA. Both protein and mRNA Cas9 formats require no plasmid manipulation and so are amenable to high throughput and multiplex genome-wide cell engineering approaches.

Features of the Precision gRNA Synthesis Kit include:
• Fast assembly and synthesis of any gRNA target in as little as four hours including template assembly
• High yield (>10 ug) and concentration (>200 ng/uL) of gRNA

How to obtain a gRNA sequence
Genome editing with CRISPR technology requires a noncoding guide RNA (gRNA) in order to cleave genomic DNA at a target sequence of interest. The gRNA has two molecular components: a target-specific CRISPR RNA (crRNA) and a trans-activating crRNA (tracrRNA) that have been combined into one transcript. The target sequence (20 bases) must be immediately upstream of a PAM motif (NGG) which allows the Cas9 to initiate binding. The PAM is only on the target DNA and not part of the target specific CRISPR sequence. The gRNA and the PAM motif guide the Cas9 nuclease to the target genomic sequence to form a complex and create a double-stranded blunt DNA break (DSB) three nucleotides upstream from the PAM site.

Use our CRISPR Search and Design Tool to search our database of >600,000 gRNA sequences specific to every gene in the human and mouse genomes. Invitrogen predesigned gRNAs are optimized for gene knockout and typically target the first three transcribed exons per gene. Search results include recommendations based on minimizing potential off-target effects, potential binding sites, and exon maps with gRNA locations. This tool can also be used to analyze any sequence of interest to design unique CRISPR sequences.

How to make gRNA
Once gRNA sequences have been selected, choose from three options for making gRNA:
1. TrueGuide synthetic guide RNA—choose from our catalog of predesigned gRNAs or upload your sequence to our TrueGuide gRNA Ordering Tool
2. Precision gRNA Synthesis Kit (this page)—for transfection-ready gRNA in as little as four hours including template assembly
3. Genome Engineering Services—save time and effort and have our custom services team design, synthesize, and purify in vitro transcribed (IVT) gRNA sequences for you. To obtain a services quotation, or to order, please contact our Custom Services department at 1-800-955-6288 x45682 or gemservices@thermofisher.com.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form
Liquid
Format
Kit
Reaction Speed
Fast
Technique
CRISPR-Cas9
Product Type
gRNA Synthesis Kit
Promoter
T7
Shipping Condition
Approved for shipment at Room Temperature or on Wet or Dry Ice
Sufficient For
25 Reactions
Yield
>10 μg at >200 ng/μL of gRNA
Concentration
200 ng/μL
Final Product Type
RNA
No. of Reactions
25 Reactions
Starting Material
Oligos (DNA)

Contents & Storage

• gRNA Prep Kit, store at -5° to -30°C
• gRNA Cleanup Kit, store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text