Applied Biosystems™

VetMAX™ Xeno™ Internal Positive Control RNA

Catalog number: A29761
Have Questions?
Applied Biosystems™

VetMAX™ Xeno™ Internal Positive Control RNA

Catalog number: A29761
Have Questions?
Catalog Number
A29761
Unit Size
500 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A29761500 reactions
-
A29763100 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
VetMAX™ Xeno™ Internal Positive Control (IPC) RNA serves as an internal positive control for the RNA purification process and helps monitor for the presence of PCR inhibitors in animal health molecular detection workflows. This is of particular importance when working with sample types that have a high level of inhibitors such as oral fluids. Xeno IPC RNA is introduced at the nucleic acid isolation/preparation step and carried through the animal health PCR workflow.

Features of VetMAX Xeno IPC RNA include:
• Provides confidence that PCR test results are accurate and actionable
• Easily integrates into any workflow
• Greatly reduces the likelihood of false negatives

VetMAX Xeno IPC RNA comes in a concentration of 10,000 copies/μL and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with a Xeno internal positive control assay, our proprietary design offers a verification layer to help ensure the qPCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.

Xeno IPC RNA is a component of our licensed VetMAX Gold detection kits, is recommended in the American Association of Veterinary Diagnosticians guidelines, and has been successfully benchmarked against millions of genomes including those relevant to animal health. Xeno IPC DNA is also available (Cat. No. A29764).
For Laboratory Use Including Veterinary and Environmental Uses.

Specifications

Format
Kit
For Use With (Application)
Detection and Identification of Animal Disease
Includes
Internal Positive Control
No. of Reactions
500
Product Line
VetMAX™, Xeno™
Product Type
Internal Positive Control RNA
Shipping Condition
Dry Ice
Species
Not Species-Specific

Contents & Storage

  • 2 x 1250 μL Nucleic Acid Dilution Solution. Store at -5°C to -30°C.
  • 1 x 1250 μL Xeno RNA Control (10,000 copies/μL). Store at -5°C to -30°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text