VetMAX™ Xeno™ Internal Positive Control RNA
VetMAX™ Xeno™ Internal Positive Control RNA
Applied Biosystems™

VetMAX™ Xeno™ Internal Positive Control RNA

VetMAX™ Xeno™ Internal Positive Control (IPC) RNA serves as an internal positive control for the RNA purification process and helpsRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
A29763100
A29761500
Catalog number A29763
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Add to cart
No. of Reactions:
100
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Add to cart
VetMAX™ Xeno™ Internal Positive Control (IPC) RNA serves as an internal positive control for the RNA purification process and helps monitor for the presence of PCR inhibitors in animal health molecular detection workflows. This is of particular importance when working with sample types that have a high level of inhibitors such as oral fluids. Xeno IPC RNA is introduced at the nucleic acid isolation/preparation step and carried through the animal health PCR workflow.

Features of VetMAX Xeno IPC RNA include:
• Provides confidence that PCR test results are accurate and actionable
• Easily integrates into any workflow
• Greatly reduces the likelihood of false negatives

VetMAX Xeno IPC RNA comes in a concentration of 10,000 copies/μL and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with a Xeno internal positive control assay, our proprietary design offers a verification layer to help ensure the qPCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.

Xeno IPC RNA is a component of our licensed VetMAX Gold detection kits, is recommended in the American Association of Veterinary Diagnosticians guidelines, and has been successfully benchmarked against millions of genomes including those relevant to animal health. Xeno IPC DNA is also available (Cat. No. A29764).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Detection and Identification of Animal Disease
IncludesInternal Positive Control
No. of Reactions100
Product LineVetMAX™, Xeno™
Product TypeInternal Positive Control RNA
Quantity100 reactions
Shipping ConditionDry Ice
SpeciesNot Species-Specific
FormatKit
Unit SizeEach
Contents & Storage
  • 1 x 1250 μL Nucleic Acid Dilution Solution. Store at -5°C to -30°C.
  • 1 x 250 μL Xeno RNA Control (10,000 copies/μL). Store at -5°C to -30°C.

Share catalog number, name or link