Alexa Fluor™ 405 NHS Ester (Succinimidyl Ester)
Alexa Fluor™ 405 NHS Ester (Succinimidyl Ester)
Invitrogen™

Alexa Fluor™ 405 NHS Ester (Succinimidyl Ester)

Alexa Fluor™ 405 is a blue-fluorescent dye optimal for use with the 405 nm violet laser. Used for stable signalRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A300001 mg
A30100
also known as A-30100
5 mg
Catalog number A30000
Price (USD)
471.00
Each
-
Add to cart
Quantity:
1 mg
Recurring order eligible. Learn more »
Price (USD)
471.00
Each
Add to cart
Ask our AI about this Product

Alexa Fluor™ 405 is a blue-fluorescent dye optimal for use with the 405 nm violet laser. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 405 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 405 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

The NHS ester (or succinimidyl ester) of Alexa Fluor™ 405 is the most popular tool for conjugating this dye to a protein or antibody. NHS esters can be used to label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting Alexa Fluor™ conjugate will exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ NHS ester:
Fluorophore label: Alexa Fluor™ 405 dye
Reactive group: NHS ester
Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
Ex/Em of the conjugate: 400/424 nm
Extinction coefficient: 35,000 cm-1M-1
Spectrally similar dyes: Pacific Blue
Molecular weight: 1028.3

Typical Conjugation Reaction
You can conjugate amine-reactive reagents with virtually any protein or peptide (the provided protocol is optimized for IgG antibodies). You can scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The Alexa Fluor™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO) (D12345), and the reaction is carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification
Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
Detection MethodFluorescence
Emission421 nm
Excitation401 nm
Flow Cytometer Laser Lines405 nm
Label or DyeAlexa Fluor™ 405
Product TypeDye
Quantity1 mg
Reactive MoietyActive Ester, Succinimidyl Ester
Shipping ConditionRoom Temperature
SolubilityDMSO (Dimethylsulfoxide)
Label TypeAlexa Fluor
Product LineAlexa Fluor™
Unit SizeEach
Contents & Storage
Contains 1 vial (1 mg). Store in freezer (-5 to -30°C) and protect from light.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

I am labeling a protein with Alexa Fluor 488 SDP ester. The manual recommends using a sodium bicarbonate buffer at pH 8.3. Can I use a different buffer instead?

Yes. The important thing is to use a buffered solution with a pH between 8.0 and 8.5. Do not use Tris buffer, which has amine groups. Most other buffers will work fine in that pH range. This is also true for other amine-reactive dyes, such as succinimidyl (NHS) esters or TFP esters.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am not going to use all of my Alexa Fluor succinimidyl ester reactive dye. Can I just make it up in DMSO and store aliquots at -20 degrees C?

This is not recommended. Any trace amounts of water in the DMSO can promote spontaneous hydrolysis over time. Even if using anhydrous DMSO, DMSO is hygroscopic; it readily absorbs moisture from the atmosphere over time. A better alternative is to dissolve the reactive dye in a volatile solvent, make smaller aliquots and then evaporate off the solvent using a vacuum pump. The smaller aliquots of solid reactive dye should then be stored frozen, desiccated and protected from light. Contact Technical Support by sending an email to techsupport@thermofisher.com for the recommended volatile solvent.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Is Alexa Fluor 405 dye the best dye for imaging with a 405 nm laser?

The "best" dye also depends on the emission filter specifications. Both Alexa Fluor 405 dye and Alexa Fluor 430 dye will be efficiently excited at 405 nm.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Fluorescence spectra

Fluorescence spectra

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3184336Certificate of AnalysisJun 29, 2025A30000
2983187Certificate of AnalysisSep 28, 2024A30000
2884573Certificate of AnalysisMar 15, 2024A30000
2833458Certificate of AnalysisDec 01, 2023A30100
2734940Certificate of AnalysisJul 17, 2023A30100
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (17)

Citations & References
Abstract
3D multicolor super-resolution imaging offers improved accuracy in neuron tracing.
Authors:Lakadamyali M, Babcock H, Bates M, Zhuang X, Lichtman J,
Journal:PLoS One
PubMed ID:22292051
'The connectivity among neurons holds the key to understanding brain function. Mapping neural connectivity in brain circuits requires imaging techniques with high spatial resolution to facilitate neuron tracing and high molecular specificity to mark different cellular and molecular populations. Here, we tested a three-dimensional (3D), multicolor super-resolution imaging method, stochastic ... More
Preparation of photoswitchable labeled antibodies for STORM imaging.
Authors:Bates M, Jones SA, Zhuang X,
Journal:
PubMed ID:23734027
'Stochastic optical reconstruction microscopy (STORM) is a method for superresolution fluorescence imaging based on the high accuracy localization of individual fluorophores. It uses optically switchable fluorophores: molecules that can be switched between a nonfluorescent and a fluorescent state by exposure to light. Many synthetic fluorescent dye molecules show photoswitchable fluorescence ... More
CD8 T cell competition for dendritic cells in vivo is an early event in activation.
Authors:Willis RA, Kappler JW, Marrack PC
Journal:Proc Natl Acad Sci U S A
PubMed ID:16880405
'T cell responses against an antigen are often focused on a small fraction of potentially immunogenic determinants, a phenomenon known as immunodominance. Immunodominance can be established at several stages of antigen presentation, including antigen processing, binding of peptides to MHC, and competition between T cells for dendritic cells (DCs). The ... More
Fast, three-dimensional super-resolution imaging of live cells.
Authors:Jones SA, Shim SH, He J, Zhuang X,
Journal:Nat Methods
PubMed ID:21552254
'We report super-resolution fluorescence imaging of live cells with high spatiotemporal resolution using stochastic optical reconstruction microscopy (STORM). By labeling proteins either directly or via SNAP tags with photoswitchable dyes, we obtained two-dimensional (2D) and 3D super-resolution images of living cells, using clathrin-coated pits and the transferrin cargo as model ... More
Superresolution imaging of chemical synapses in the brain.
Authors:Dani A, Huang B, Bergan J, Dulac C, Zhuang X,
Journal:Neuron
PubMed ID:21144999
'Determination of the molecular architecture of synapses requires nanoscopic image resolution and specific molecular recognition, a task that has so far defied many conventional imaging approaches. Here, we present a superresolution fluorescence imaging method to visualize the molecular architecture of synapses in the brain. Using multicolor, three-dimensional stochastic optical reconstruction ... More
17 total citations

Share catalog number, name or link

1x1 image pixel for data collection