Invitrogen™

Alexa Fluor™ 488 5-SDP Ester (Alexa Fluor™ 488 Sulfodichlorophenol Ester)

Catalog number: A30052
Have Questions?
Invitrogen™

Alexa Fluor™ 488 5-SDP Ester (Alexa Fluor™ 488 Sulfodichlorophenol Ester)

Catalog number: A30052
Have Questions?
Catalog Number
A30052
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A300521 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The SDP (sulfodicholorphenol) ester is currently the most hydrolytically stable amine-reactive moiety available from Invitrogen. Conjugates produced with the Alexa Fluor™ 488 5-SDP ester produce the same strong amide bond between the dye and the compound of interest as succinimidyl (SE or NHS) and tetrafluorophenyl (TFP) esters, but with the improved stability in water and buffers, the SDP ester can potentially offer increased control and consistency in these reactions compared to the NHS and TFP esters.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
1 mg
Product Type
Sulfodichlorophenol Ester
Chemical Reactivity
Amine
Shipping Condition
Room Temperature
Reactive Moiety
Active Ester, SDP Ester
Label or Dye
Alexa Fluor™ 488
Label Type
Alexa Fluor
Product Line
Alexa Fluor™

Contents & Storage

Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text