Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 µg, 10 µg and 20 µg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 µL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase.
This CAMK1G (CaMKI gamma) recombinant human protein (full length) was expressed in insect cells.
CAMK1G (CaMKI gamma) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service.
• Validated for use with the Z'-LYTE™ assay
• Available in multiple pack sizes for convenient use
• Also available in our SelectScreen Kinase Profiling Service
Protein Form: Full Length, Recombinant, Active
CAMK1G serine/threonine kinase is a member of CAMK family and is a novel membrane-anchored neuronal Ca2+ calmodulin-dependent protein kinase. Full activation of CAMK1G requires both Ca2+ and calmodulin and phosphorylation by CAMKK. CAMK1G transcripts are most abundant in neurons, with the highest levels in limited nuclei such as the central nucleus of the amygdala (CeA) and the ventromedial hypothalamus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium/calmodulin-dependent protein kinase IG; Calcium/calmodulin-dependent protein kinase type 1G; CaM kinase 1G; CaM kinase I gamma; CaM kinase IG; caM-KI gamma; CaMK-like CREB kinase III; caMKI gamma; caMKIG; CLICK III; RP1-272L16.2
Gene Aliases: CAMK1G; CLICK3; CLICKIII; dJ272L16.1; VWS1
UniProt ID: (Human) Q96NX5
Entrez Gene ID: (Human) 57172
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support