1x1 image pixel for data collection
Ion 520™ & Ion 530™ ExT Kit-Chef
Ion 520™ & Ion 530™ ExT Kit-Chef
Ion Torrent™
Ion 520™ & Ion 530™ ExT Kit-Chef
The Ion 520™ & Ion 530™ ExT Kit-Chef enables robust, automated, and fast template preparation and sequencing of 400 toRead more
Have Questions?
Catalog NumberQuantity
A306704 reactions
Catalog number A30670
Price (USD)
7,630.00
Each
Add to cart
Quantity:
4 reactions
Price (USD)
7,630.00
Each
Add to cart
The Ion 520™ & Ion 530™ ExT Kit-Chef enables robust, automated, and fast template preparation and sequencing of 400 to 600 base-read libraries using the Ion Chef™ System and the Ion S5™ and Ion S5™ XL Sequencing Systems. The Ion 520 & Ion 530 ExT Kit-Chef contains pre-packaged single-use template and sequencing reagent cartridges with integrated sample tracking, delivering an automated workflow for sequencing of 4 loaded chips with complete run traceability.
The Ion 520 & Ion 530 ExT Kit-Chef and Ion S5/Ion S5 XL workflow:
• Provides template preparation and chip loading reagents for 400 to 600 base-read libraries
• Simplifies your research with ready-to-use, disposable reagent cartridges for templating and sequencing
• Enables fast and simple sequencing instrument set-up, with less than 15 minutes hands-on time
• Leverages the latest advancements in data quality and accuracy
• Maximizes flexibility with compatibility for both the Ion 520™ and Ion 530™ chips
The Ion S5/Ion S5 XL sequencers, along with the Ion Chef System, offer reproducible sequencing workflows, with minimal hands-on time. Choose to pair the Ion 520 & Ion 530 ExT Kit with the Ion 520 Chip Kit (up to 4 million reads per chip) or Ion 530 Chip Kit (up to 12 million reads), allowing for flexibility of total throughput. For de novo research applications, add the Ion S5™ ExT Calibration Standard to prepared libraries for greater base-calling accuracy of 400 to 600 bp fragments.
Notes:
This kit is only compatible with the Ion S5 and Ion S5 XL Systems.
Use this kit with Torrent Suite™ Software 5.2 or higher.
Specifications
For Use With (Application)Resequencing
For Use With (Equipment)Ion GeneStudio S5 System, Ion S5™ XL System
Product TypeChef Kit
Quantity4 reactions
Throughput400 to 500 Base-Read Libraries
Product LineIon Torrent™
Software TypeTorrent Suite Software 5.2
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection