HID Real-Time PCR Analysis Software v1.3
For use with QuantStudio 5 firmware version 1.5.1
Product Image
Applied Biosystems™
HID Real-Time PCR Analysis Software v1.3
HID Real-Time PCR Analysis Software is a tool that helps enable forensic laboratories to optimize their quantification workflow and enhanceRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A311525 Licenses
A311501 License
Catalog number A31152
Price (USD)
53,100.00
Each
-
Add to cart
Quantity:
5 Licenses
Price (USD)
53,100.00
Each
Add to cart
HID Real-Time PCR Analysis Software is a tool that helps enable forensic laboratories to optimize their quantification workflow and enhance their downstream decision-making process. The software has the following workflow features:
• Virtual standard curve functionality
• Customizable templates
• Advanced data analysis capabilities including quality flags
• STR sample normalization (dilution) and reaction setup tool
HID Real-Time PCR Analysis Software v1.3 is designed for use with the Applied Biosystems 7500 and QuantStudio 5 real-time PCR systems. It offers a set of features beyond instrument controls. HID Real-Time PCR Analysis Software v1.3 streamlines the experimental setup, supports customizable workflow templates, and enhances the data analysis and review process. It is validated for use on the Windows™ 7 platform and is may be used with all available HID Quantifiler kits.
This product includes an HID Real-Time PCR Analysis Software v1.3 DVD and 1 user license. Documentation is not included.
Specifications
For Use With (Equipment)Applied Biosystems 7500, QuantStudio 5 Real-Time PCR System
Product TypeHID Real-Time PCR Analysis Software V1.3
Quantity5 Licenses
Shipping ConditionRoom Temperature
Software TypeReal-Time PCR Analysis
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link