Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

PureLink™ Expi Endotoxin-Free Maxi Plasmid Purification Kit

Catalog number: A31217
Have Questions?
Invitrogen™

PureLink™ Expi Endotoxin-Free Maxi Plasmid Purification Kit

Catalog number: A31217
Have Questions?
Catalog Number
A31217
Unit Size
10 preps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 preps (sample size)
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25 preps
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A3121710 preps
-
A330734 preps (sample size)
-
A3123125 preps
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The PureLink Expi Endotoxin-Free Maxi Plasmid Purification Kit enables quick and simple isolation of endotoxin-free plasmid DNA using an enhanced anion exchange membrane. In as little as 90 minutes, purify up to 1.5 mg of high-quality plasmid DNA that is suitable for advanced transfection applications such as transfection of sensitive cells lines or genome engineering.

Advantages of the PureLink Expi Endotoxin-Free Maxi Plasmid Purification Kit include:
• Simple and fast protocol—purification typically takes 90 minutes
• High yield—isolate up to 1.5 mg of high quality plasmid DNA
• Endotoxin-free plasmid—less than 0.1 EU/μg, ideal for sensitive applications
• High purity—ultralow levels of protein, RNA, and genomic DNA contamination
• Compatibility—suitable for use with ExpiCHO and Expi293 expression systems
• Endotoxin-free components— elution buffer, water, and plasmid storage tubes that are free of endotoxin

Simple and fast protocol
Unlike other anion-exchange-based endotoxin-free plasmid purification systems, the PureLink Expi Endotoxin-Free Maxi Plasmid Purification Kit utilizes centrifugation- or vacuum-assisted filtration for lysate clarification and subsequent plasmid DNA purification. There is no waiting for gravity-flow columns and the entire purification typically takes 90 minutes (including the final precipitation step).

Endotoxin-free plasmid
Purify endotoxin-free (less than 0.1 EU/μg) plasmid that is ideal for sensitive applications such as transfection of primary cells, genome engineering, and research on gene therapy or plasmid vaccines in vivo.

Compatible with ExpiCHO and Expi293 expression systems
Plasmid purified using the PureLink Expi Endotoxin-Free Maxi Plasmid Purification Kit is fully compatible with the Gibco ExpiCHO and Expi293 transient mammalian expression systems.

Superior quality with endotoxin-free components
Rest assured that the kit is of high quality and has additional endotoxin-free elution buffer, water, and plasmid storage tubes for added peace of mind.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Spin Column
Isolation Technology
Anion Exchange Resin
Sample Type
Bacteria
Scale
Maxi
Expression System
Mammalian
For Use With (Application)
Cloning, PCR
High-throughput Compatibility
Manual Protocols
No. of Reactions
10 Preps
Prep Scale
> 1 mg (Large-Scale) Plasmid DNA, > 200 μg (Large-Scale) Plasmid DNA
Product Line
PureLink™
Product Type
Plasmid Purification Kit
Quantity
10 Preps
Shipping Condition
Room Temperature
Target
Plasmid DNA
Test Time
90 min.

Contents & Storage

• Resuspension buffer
• Lysis indicator
• RNase A
• Lysis buffer
• Precipitation buffer
• Endotoxin removal buffer
• Wash buffer
• Elution buffer
• TE buffer
• Expi Maxi filtration column
• Expi Maxi DNA binding column
• 50 mL conical tubes

Endotoxin-free components: elution buffer, water, plasmid storage tubes

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text