The DS-37 Matrix Standard Kit (Dye Set J6-T) is used to perform the spectral calibration required to analyze 6-FAM, VIC,Read more
Have Questions?
Catalog number A31234
Price (USD)/ 8 runs
319.00
Add to cart
Price (USD)/ 8 runs
319.00
Add to cart
DS37 Matrix Standard Kit (Dye Set J6-T)
Catalog numberA31234
Price (USD)/ 8 runs
319.00
Add to cart
The DS-37 Matrix Standard Kit (Dye Set J6-T) is used to perform the spectral calibration required to analyze 6-FAM, VIC, TED, SID, TAZ, and LIZ dye-labeled DNA fragments on the Applied Biosystems SeqStudio, 3130, and 3500 series systems. The data collection software for these instruments uses the multi-component matrix to automatically analyze the 6 differently colored fluorescent dye-labeled samples in a single capillary. The DS-37 Matrix Standard Kit (Dye Set J6-T) contains 6 specific sizes of a unique fluorescent dye label.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Software TypeData Collection Software
Starting MaterialDNA
For Use With (Equipment)SeqStudio, 3130 and 3500 Series System