Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermal Accuracy | ±0.25°C (35°C to 99.9°C) |
High-throughput Compatibility | High-throughput Compatible |
Thermal Uniformity | <0.5°C (20 sec. after reaching 95°C) |
Dimensions | 12.5 x 7.1 x 5.3 in. (31.7 x 17.9 x 13.3 cm) |
Capacity | 96-well, 0.2 mL, full or half-skirted plate |
Voltage | 100/240 V |
Display Type | No Display |
Product Type | Thermal Cycler |
Warranty | Limited Warranty |
Thermal Range | 0°C to 105°C |
User Interface | USB, Wi-Fi, Ethernet |
Wattage | 600 W |
Max. Ramp Rate | 3.5°C/sec. (Block), 1.8°C/sec. (Sample) |
Description | Includes a 3-meter connector cable |
Weight | 13.2 lb. (6 kg) |
Block Configurations | 96-well Isothermal Block |
Unit Size | 1 each |
Thermal Accuracy | ±0.25°C (35°C to 99.9°C) |
High-throughput Compatibility | High-throughput Compatible |
Thermal Uniformity | <0.5°C (20 sec. after reaching 95°C) |
Dimensions | 12.5 x 7.1 x 5.3 in. (31.7 x 17.9 x 13.3 cm) |
Capacity | 96-well, 0.2 mL, full or half-skirted plate |
Voltage | 100/240 V |
Display Type | No Display |
Product Type | Thermal Cycler |
Warranty | Limited Warranty |
Thermal Range | 0°C to 105°C |
User Interface | USB, Wi-Fi, Ethernet |
Wattage | 600 W |
Max. Ramp Rate | 3.5°C/sec. (Block), 1.8°C/sec. (Sample) |
Description | Includes a 1-meter connector cable |
Weight | 13.2 lb. (6 kg) |
Block Configurations | 96-well Isothermal Block |
Unit Size | 1 each |
Thermal Accuracy | ±0.25°C (35°C to 99.9°C) |
High-throughput Compatibility | High-throughput Compatible |
Thermal Uniformity | <0.5°C (20 sec. after reaching 95°C) |
Dimensions | 12.5 x 7.1 x 5.3 in. (31.7 x 17.9 x 13.3 cm) |
Capacity | 96 x 0.2 mL, full or half-skirted plates |
Voltage | 100/240 V |
Display Type | No Display |
Product Type | Thermal Cycler |
Warranty | Limited Warranty |
Thermal Range | 4°C to 105°C |
User Interface | USB, Wi-Fi, Ethernet |
Wattage | 600 W |
Max. Ramp Rate | 3.5°C/sec. (Block), 1.8°C/sec. (Sample) |
Description | Includes a 10-cm connector cable |
Weight | 13.2 lb. (6 kg) |
Block Configurations | 96-well Isothermal Block |
Unit Size | 1 each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A31490 | 1 each | Includes a 3-meter connector cable | 18,910.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A31489 | 1 each | Includes a 1-meter connector cable | 15,325.65 Online price 18,800.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A31491 | 1 each | Includes a 10-cm connector cable | 15,692.65 Online price 19,250.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Space-saving design with our smallest thermal cycler footprint
• Flexible modular design helps save space on a robotic deck
• Separate control panel module provides flexibility when integrating on robotic deck
• Choice of 10-cm, 1-m, and 3-m cables allows flexibility to locate control module under thermal cycler, under deck, or up to 3 meters away
Automated lid and cycler features
• Automated lid compatible with programming for easy, hands-free operation with liquid handler or plate stacker
• Heated cover slides forward to cover plate nest, helping ensure thermal contact and eliminate evaporation
• Compatible with full-skirt or half-skirt plates
Fully integrate your automated system with our easy-to-install software
• Integrate with robot via SiLA Rapid Integration standard–compatible software and prewritten application programming interfaces (APIs) for major robotic platforms
• Test instrument using desktop software before buying robot
• Enable ATC to cycle like old instrument with Applied Biosystems 9700 system simulation modes
Use the ATC as a stand-alone PCR thermal cycler
Flexible software can be connected to your robot's software interface or used to control the ATC as a stand-alone instrument. Test your instrument using the ATC desktop application before integrating it into your robotic automation system.
Compatible plastics for optimal performance
MicroAmp EnduraPlate Optical 96-Well Full-Skirted Plates are designed for use with the ATC.
• Full-skirted polycarbonate frame constructed to ANSI/SBS standards for compatibility with robotic handling and plate stacking
• Three-way barcodes on east (A12-H12), south (H1-H12), and west (A1-A12) sides to improve plate reading efficiency
• Easy-to-read black well-identification text to minimize pipetting errors
• Available in multiple colors for easy organization of laboratory assays
• Certified DNA-, RNase-, and PCR inhibitor-free
Award-winning service and support
• Help reduce downtime with rapid exchange support
• Have our engineers perform temperature qualification at your site
Applied Biosystems PCR plastics
Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for any experiment.