Informational:

***ATTENTION***

*** ITEM DISCONTINUATION***



These products are being replaced with Premium (FBS) performance-based products. The Premium product specifications are improved and product stability, performance, manufacturing, storage, and transport processes are unchanged.

New and improved replacement available, Buy now›
Gibco™

Fetal Bovine Serum, qualified, One Shot™ format, United States

Catalog number: A3160502
Have Questions?
Gibco™

Fetal Bovine Serum, qualified, One Shot™ format, United States

Catalog number: A3160502
Have Questions?
Catalog Number
A3160502
Unit Size
10 x 50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A316050210 x 50 mL
-
A316050150 mL
-
26140079500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

Gibco Sera: Trusted Quality for Consistent Performance

Gibco Sera Category: Value Plus (Performance)

  • Use for general cell culture with common cell lines
  • Low endotoxin and hemoglobin levels
  • Endotoxin level: <10 EU/mL
  • Hemoglobin level: <25 mg/dL
  • Origin: United States

Gibco Serum Delivers

  • ISO 13485 certified, processed in FDA registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

Specifications

Description
Fetal Bovine Serum, qualified, One Shot™ format, United States
Age
Fetal
Species
Bovine
Country of Origin
United States
Form
Liquid
Format
One Shot
Product Type
Fetal Bovine Serum
Quantity
10 x 50 mL
Serum Treatment
Standard (Sterile-filtered)
Sterility
Sterile
Sterilization Method
Triple-filtered, 0.1 μm
Endotoxin Concentration
≤10 EU/mL
Green Features
Fewer resources used and less waste
Hemoglobin Concentration
≤25 mg/dL
Purity or Quality Grade
Qualified
Product Line
One Shot™
Shipping Condition
Frozen

Contents & Storage

Storage conditions: ≤-10°C
Shipping conditions: Frozen

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text