Vitronectin (VTN-N) Recombinant Human Protein, Truncated
Taking your work to the clinic? Check out CTS™ VTN-N »
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Gibco™
Vitronectin (VTN-N) Recombinant Human Protein, Truncated
Vitronectin (VTN-N) is a recombinant human protein that provides a defined surface for feeder-free culture of human pluripotent stem cellsRead more
Have Questions?
Change view
Catalog Number
Quantity
A31804
10 mL = 5 mg
A14700
1 mL = 0.5 mg
2 Options
Catalog number A31804
Price (USD)
900.00
Each
-
Add to cart
Quantity:
10 mL = 5 mg
Price (USD)
900.00
Each
Add to cart
Ask our AI about this Product
Vitronectin (VTN-N) is a recombinant human protein that provides a defined surface for feeder-free culture of human pluripotent stem cells (PSCs). When used with Essential 8™ Medium (A1517001), vitronectin (VTN-N) has been proven to maintain pluripotency and normal growth characteristics in multiple PSC lines. In addition, vitronectin (VTN-N) has been shown to support PSC growth for >50 passages without any signs of karyotypic abnormalities and to maintain the ability of PSCs to differentiate into all three germ line lineages. As published by Guokai Chen et al. (1) in the laboratory of James Thomson, the VTN-N variant of vitronectin supports human pluripotent stem cell attachment and survival better than wild-type vitronectin when used in conjunction with Essential 8™ Medium.
Vitronectin (VTN-N) is:
• Optimized—designed in the laboratory of James Thomson for use with Essential 8™ Medium • Defined—for reduced variability in your PSC cultures • Cost effective—for economical and scalable PSC culture
Optimized and Defined Surface Unlike standard basement membrane extracts (BMEs), vitronectin (VTN-N) was designed in the laboratory of James Thomson for use with Essential 8™ Medium and has been shown to support human pluripotent stem cell attachment and survival better than wild type vitronectin. Vitronectin (VTN-N) provides an optimal environment for feeder-free pluripotent stem cell culture.
Reduce Variability Vitronectin (VTN-N) is a defined, recombinant human protein that reduces variability in your PSC cultures compared to human-plasma derived vitronectin and standard basement membrane extracts (BMEs).
Cost Effective When used with Essential 8™ Medium, vitronectin (VTN-N) provides a cost effective, defined system for feeder free culture of human pluripotent stem cells (PSCs).
Commercialized in Partnership with Cellular Dynamics International.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration500 μg⁄ml
Expression SystemE. coli
Protein FormRecombinant
Protein SubtypeVitronectin
Purity or Quality GradeCell Culture Grade, Endotoxin-Screened
Quantity10 mL = 5 mg
Research CategoryStem Cell Research
Shipping ConditionDry Ice
Volume (Metric)10 mL
For Use With (Application)Stem Cell Culture
FormLiquid
RecombinantRecombinant
SpeciesHuman
Unit SizeEach
Contents & Storage
Store at -80°C and protect from light.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.
Frequently asked questions (FAQs)
What substrate should I use with the PSC Definitive Endoderm Induction Kit?
We recommend using Vitronectin (VTN-N) Recombinant Human Protein, Truncated (Cat. No. A14700), but Geltrex LDEV-Free hESC-qualified Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1413301 or Cat. No. A1413302) can also be used.
Can pluripotent stem cells (PSCs) cryopreserved in a different culture condition be thawed and subsequently cultured in Gibco Essential 8 Medium and on vitronectin (VTN-N)?
Yes. PSCs cryopreserved from cultures of mTeSR Medium and BD Matrigel Basement Membrane Matrix may be thawed into Gibco Essential 8 Medium and plated on VTN-N. Certain lines may benefit from thawing into the medium and substrate they were growing in at the time of cryopreservation. Then at the next passage, use EDTA to passage the cells into Gibco Essential 8 Medium and VTN-N.
Can I use enzymes such as dispase and collagenase for passaging cells cultured in Gibco Essential 8 Medium and on VTN-N?
Enzymes such as dispase and collagenase do not work well with cells cultured in Gibco Essential 8 Medium on VTN-N. Use of these enzymes for passaging cells results in compromised viability and attachment.
What will my cells look like growing in Gibco Essential 8 Medium and on Gibco Vitronectin (VTN-N)?
You should expect to see normal pluripotent stem cell (PSC) morphology. The expected morphology of PSCs is demonstrated specifically by tightly packed colonies with defined borders and a high nucleus-to-cytoplasm ratio.