Ion Torrent™

Tag Sequencing Barcode Set 1-24

Catalog number: A31830
Have Questions?
Ion Torrent™

Tag Sequencing Barcode Set 1-24

Catalog number: A31830
Have Questions?
Catalog Number
A31830
Unit Size
1 set
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A318301 set
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Tag Sequencing Barcode Set 1-24 provides a set of 24 unique barcode adapters specifically designed for optimal performance in Oncomine™ cfDNA assays. The barcode adapters are compatible with the Ion S5™, Ion PGM™, and Ion Proton™ sequencing systems. When used in combination with kits utilizing tag sequencing technology, this barcode set enables users to pool up to 24 amplicon libraries and to conduct multiplexed next-generation sequencing (NGS) analysis, reducing the sequencing cost per sample.

Key product features:
• Enables multiplexing of numerous amplicon library samples on a single sequencing chip with the use of robust molecular barcodes
• Both sequence-optimized and flow-optimized for equal representation of all barcodes in a pool and more economical multiplexed sequencing runs
• Compatible with library construction kits utilizing tag sequencing technology

Multiplexing with barcode adapters enables higher throughput
Multiplexing with barcoded libraries leads to more cost-effective runs by permitting up to 24 samples per run (or more if combined with Tag Sequencing BC Set 25-48, Cat. No. A31847), significantly decreasing the cost and handling requirements of sequencing. Each barcode was individually tested to minimize representation bias.

Sequence-optimized and flow-optimized for increased performance and efficiency
Barcode adapters contain sequences that are optimized to provide equal representation of all barcodes in a pool; they require a minimal number of flows to interrogate resulting in more economical multiplexed sequencing runs.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Application)
Next-Generation Sequencing (NGS)
No. of Reactions
10 Reactions
Primer
Custom Primer
Shipping Condition
Dry Ice
Form
Liquid

Contents & Storage

10 μL each Tag Sequencing Barcodes 01 thru 24, 10 reactions/barcode

Store in freezer (-5°C to -30°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text