Ion Torrent™

Tag Sequencing Barcode Set 25-48

Número de catálogo: A31847
Ion Torrent™

Tag Sequencing Barcode Set 25-48

Número de catálogo: A31847
Número de catálogo
A31847
Tamaño de la unidad
1 set
Precio (MXN)
Número de catálogoTamaño de la unidadPrecio (MXN)
A318471 setContáctenos ›
Descripción general del producto
Figuras
Videos
Recomendaciones
Recomendaciones
Documentos
Preguntas frecuentes
Citas y referencias
Información adicional
Recomendaciones
The Tag Sequencing Barcode Set 25-48 provides a set of 24 unique barcode adapters specifically designed for optimal performance in Oncomine™ cfDNA assays. The barcode adapters are compatible with the Ion S5™, Ion PGM™, and Ion Proton™ sequencing systems. When used in combination with kits utilizing tag sequencing technology, this barcode set enables users to pool up to 24 amplicon libraries and to conduct multiplexed next-generation sequencing (NGS) analysis, reducing the sequencing cost per sample.

Key product features:
• Enables multiplexing of numerous amplicon library samples on a single sequencing chip with the use of robust molecular barcodes
• Both sequence-optimized and flow-optimized for equal representation of all barcodes in a pool and more economical multiplexed sequencing runs
• Compatible with library construction kits utilizing tag sequencing technology

Multiplexing with barcode adapters enables higher throughput
Multiplexing with barcoded libraries leads to more cost-effective runs by permitting up to 24 samples per run (or more if combined with Tag Sequencing BC Set 1-24, Cat. No. A31830), significantly decreasing the cost and handling requirements of sequencing. Each barcode was individually tested to minimize representation bias.

Sequence-optimized and flow-optimized for increased performance and efficiency
Barcode adapters contain sequences that are optimized to provide equal representation of all barcodes in a pool; they require a minimal number of flows to interrogate resulting in more economical multiplexed sequencing runs.

For Research Use Only. Not for use in diagnostic procedures.

Especificaciones

Tipo de producto
Oligonucleotides, Primers
Condiciones de envío
Dry Ice
Primer
Custom Primer
Cantidad
1 Set

Contenido y almacenamiento

10 μL each Tag Sequencing Barcodes 24 thru 48, 10 reactions/barcode

Store in freezer (-5 to -30°C).

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text