Informational:Informational:100% knock-in using CRISPR-Cas9—try our new TrueTag Donor DNA Kits for fluorescent or epitope gene tagging.
Invitrogen™

LentiArray™ CRISPR Negative Control Lentivirus, human, non-targeting, with GFP

Catalog number: A32063
Have Questions?
Invitrogen™

LentiArray™ CRISPR Negative Control Lentivirus, human, non-targeting, with GFP

Catalog number: A32063
Have Questions?
Catalog Number
A32063
Unit Size
100 µL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A32063100 µL
-
A328311 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Negative control gRNAs are essential in screening applications for setting thresholds for hit determination. The Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, Human Scrambled, contains a gRNA sequence with no sequence homology to any region of the human genome. This construct also expresses emGFP to provide a visual readout of successful transduction that can be used in determining Multiplicity of Infection (MOI).

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Delivery Type
Lentiviral
Format
Tube
Control Type
Negative Control
RNAi Type
gRNA
Species
Human
For Use With (Application)
Genome Editing
Product Line
LentiArray™
Product Type
gRNA
Promoter
U6
Quantity
100 mL
Shipping Condition
Dry Ice

Contents & Storage

1 x 100 μL. Store (at -68°C to -85°C.)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text