Informational:Informational:100% knock-in using CRISPR-Cas9—try our new TrueTag Donor DNA Kits for fluorescent or epitope gene tagging.
Invitrogen™

LentiArray™ Cas9 Lentivirus

Catalog number: A32069
Have Questions?
Invitrogen™

LentiArray™ Cas9 Lentivirus

Catalog number: A32069
Have Questions?
Catalog Number
A32069
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A320691 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
LentiArray Cas9 Lentivirus provides an efficient method to drive high level Cas9 nuclease expression in a wide variety of cell types. The LentiArray Cas9 Lentivirus construct includes a human codon-optimized version of Cas9 with two nuclear localization signals to facilitate efficient delivery into the nucleus. The 1-mL size is designed to support studies utilizing a co-infection approach (Cas9 lentivirus + guideRNA lentivirus delivered together) with either the LentiArray CRISPR libraries or LentiArray gRNAs against single gene targets. LentiArray Cas9 Lentivirus is provided as ready to use lentivirus at a titer of 1 x 107 TU/mL.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Delivery Type
Lentiviral
Sample Type
siRNA
Promoter
EF-1a
Transfection Technique
Lentiviral transduction
Species
Human
For Use With (Application)
CRISPR gene knockout
Product Line
LentiArray™
Product Type
Cas9 Lentivirus
Quantity
1 mL
Shipping Condition
Dry Ice

Contents & Storage

1 x 1 mL. Store at -68 to -85°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text