The Torrent Storage™ NAS Device provides network-attached storage (NAS) for Ion Torrent™ sequencing runs and increases available storage space on your Torrent Server. The NAS Device connects to the Torrent Server directly or through your local network. Torrent Storage NAS devices are installed by one of our field service engineers.
•
Save data in your lab or data center
•
Share storage with multiple Ion S5™ instruments and Torrent Servers
•
Transfer data over direct network or LAN
•
Access stored data over your network
•
Use reliable storage with disk failure tolerance
•
Expand storage as needed*
Onsite installation and configurationAfter the NAS Device is ordered, it is installed locally by a field service engineer to ensure proper configuration.
Direct file export from Torrent Suite SoftwareFiles generated from an Ion instrument are saved on the Torrent Server and can be transferred directly to the NAS Device via the Torrent Suite™ software interface. After transfer of the data, the files are available for re-analysis or archiving.
Torrent Storage NAS 110T device capacityFile type | Ion 318™ Chip | Ion 520™ Chip | Ion 530™ Chip | Ion 540™ Chip |
WELLS and BAM | 7850 | 5500 | 1800 | 450 |
BAM files only | 22000 | 22000 | 8400 | 2100 |
Note: These numbers are approximate based on 200 bp runs.
Services and support Service and support plans—comprehensive plans to maximize instrument performance and ensure availability of critical systems
Training—courses for experimental design, data analysis, troubleshooting, applications, and instrument operation
Bioinformatics—tailored support for NGS informatics execution, software use, data analysis, and downstream applications
DataSafe Solution—storage and back-up strategy, design, hardware, and support for all lab data
*Storage expansion is limited.
For Research Use Only. Not for use in diagnostic procedures.