Ion Torrent™
E.Coli DH10B Control 600 Library
Catalog number: A32470
Have Questions?
Ion Torrent™
E.Coli DH10B Control 600 Library
Catalog number: A32470
Have Questions?
Catalog Number
A32470
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A324701 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The E.coli DH10b Control 600 Library provides a templating control for use with the Ion 520™ & Ion 530™ ExT Kit- Chef and the Ion S5™ and Ion S5™ XL Sequencing Systems. The kit offers greater experimental control and empowers you to assess sample quality, help save time and reagents, and maximize productivity. Each kit contains a 600 bp E.coli DH10b control library for up to 10 template preparations.
Notes:
This kit is only compatible with the Ion S5 and Ion S5 XL Sequencing Systems.
Use this kit with the Ion 520™ & Ion 530™ ExT Kit-Chef and Torrent Suite™ Software 5.2 or higher.
Specifications
For Use With (Equipment)
Ion GeneStudio S5 System, Ion S5™ XL System
For Use With (Application)
Sequencing
Product Line
Ion Torrent™
Product Type
DH10B Control 600 Library
Quantity
1 Kit