KingFisher™ Flex Purification System with 96 Deep-Well Head
Build your own complete bundle and get personalized pricing! Learn more now.
KingFisher™ Flex Purification System with 96 Deep-Well Head
Thermo Scientific™
KingFisher™ Flex Purification System with 96 Deep-Well Head
Prepare samples efficiently from food and environmental samples with this purification system.
Have Questions?
Catalog number A32681
Price (USD)
-
Request A Quote
Ask our AI about this Product
Carry out automated total nucleic acid (DNA and/or RNA) purification and concentration of selected targets from a broad range of pre-enriched food and environmental samples with the Thermo Scientific™ KingFisher™ Flex Purification System with 96 Deep-well Head. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality in demanding high throughput workflows.
The KingFisher Flex with 96 Deep-well Head has been optimized for pathogen detection, GMO and Species Identification, and SARS-CoV-2 detection workflows.
Find a flexible solution for different types of sample processing needs. With a 96 deep-well magnet head, customers can raise the processing volume up to 5 mL, resulting in increased yield of the purified product.
  • Fully automated system yields high-speed purification of nucleic acids.
  • High throughput system processes up to 96 samples.
  • Open and flexible system lets the customer use any magnetic particle-based kit to meet the application demands.
  • Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation and modification.
  • Ready-made protocols for different type of applications are available.
Recommended for:
  • DNA isolation in Food and Environmental pathogen detection including Salmonella , Listeria spp., Listeria monocytongenes, E. coli O157:H7, and our new multiplexed assay for simultaneous detection and differentiation of Salmonella species, Typhimurium and Enteritidis
  • DNA isolation for GMO and Species Identification
  • RNA isolation SARS-CoV-2 detection in Food and Environmental samples
The instrument comes pre-installed with optimized protocols and additional protocol scripts are easily available to download from our website (here). The instrument does not require any further optimization after installation and verification.
For related information visit PCR Sample Preparation for Food Testing
For Testing of Food and Environmental Samples Only.
For Testing of Food and Environmental Samples Only.
Specifications
DescriptionKingFisher Flex 96 with Deep-well Head
For Use With (Equipment)
  • Applied Biosystems 7500 Fast Real-Time Food Safety PCR instrument
  • Applied Biosystems QuantStudio™ 5 Food Safety Real Time PCR System
  • Ion Chef™ Food Protection Instrument
  • Ion GeneStudio™ S5 Plus Food Protection System
Unit SizeEach
Group-Meat-Species-ID-kit-boxes-480x212
Have confidence in the authenticity of your food ingredients with our real-time PCR solutions
Protect your food business with our complete portfolio of PCR testing solutions for food authenticity testing. Have confidence in the products you release with assays that offer leading sensitivity far below the required limit of detection (LOD) and a rapid time-to-result that allows for faster distribution or intervention. 
Explore the complete range ›
Have questions about this product? Ask our AI assisted search.
What is the density (g/L) for RPMI 1640 Medium?
Should I be concerned about phenol red in my media when labeling my live cells with fluorescent dyes?
I understand that some media are worse than others for fluorescence imaging. How do I choose?
Do you offer media without phenol red (phenol red-free media)?
How light sensitive is RPMI 1640 media? Should I also be protecting it from LED light?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Frequently asked questions (FAQs)
The only way is to do this is by rewriting the script from where you want it to start, or start the run all over again.
There is no way to edit a BindiT protocol that has been installed on the KingFisher Flex/KingFisher DuoPrime instrument. The only way to edit the protocol is by saving the original .bdz file to a PC and then editing it in BindiT software.
Share catalog number, name or link
1x1 image pixel for data collection