Labeled 5-aminohexylacrylamido-dUTP (aha-dUTP) nucleotides are modified at the C-5 position of uridine with a unique hexylacrylamide linker, which serves asRead more
Have Questions?
Catalog number A32762
Price (USD)/ 50 μl
772.00
-
Add to cart
Price (USD)/ 50 μl
772.00
Add to cart
Alexa Fluor™ 555-aha-dUTP, 1 mM in TE buffer
Catalog numberA32762
Price (USD)/ 50 μl
772.00
-
Add to cart
Labeled 5-aminohexylacrylamido-dUTP (aha-dUTP) nucleotides are modified at the C-5 position of uridine with a unique hexylacrylamide linker, which serves as a spacer between the nucleotide and the dye. This spacer reduces interactions between the nucleotide and the dye, resulting in brighter conjugates and increased hapten accessibility for secondary detection reagents. Labeled nucleotides can be used to generate labeled nucleic acid hybridization probes for many molecular biology and molecular cytogenetics applications, including multicolor techniques.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeLabeled Nucleotide
Concentration1 mM
Label or DyeAlexa Fluor Dye
ModificationAHA (5-aminohexylacrylamido)
Shipping ConditionWet Ice
Product LineAlexa Fluor™
For Use With (Application)Molecular Biology
Quantity50 μL
Unit Size50 μl
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.