Applied Biosystems™

MagMAX™ CORE Mechanical Lysis Module

Catalog number: A32836
Have Questions?
Applied Biosystems™

MagMAX™ CORE Mechanical Lysis Module

Catalog number: A32836
Have Questions?
Catalog Number
A32836
Unit Size
100 tests
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A32836100 tests
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems MagMAX Core sample preparation system is a universal system designed to meet current and future veterinary testing needs. This modular technology is optimized for the purification of nucleic acids from a wide range of sample matrices commonly submitted for veterinary diagnostic testing. The MagMAX CORE Mechanical Lysis Module is offered as a supplement to the MagMAX CORE Nucleic Acid Purification Kit to aid in the processing of hard-to-lyse organisms like Mycobacterium avium paratuberculosis (MAP) and bovine tuberculosis in feces, milk, and environmental samples.

For a kit that contains this Mechanical Lysis Module as well as the MagMAX CORE Nucleic Acid Purification Kit, please see Cat. No. A37487.

Features include:
Convenience—a complete solution using the same CORE chemistry
Confidence—higher-quality nucleic acids from difficult-to-lyse samples, including both small (0.3 g) and large (2–5 g) volume feces samples
Flexible—purchase only what you need to lyse MAP and other hard-to-lyse organisms

The MagMAX CORE kit and the MagMAX Core Mechanical Lysis Module help make your lab more efficient through greater convenience, consistent performance, and time and cost savings.

Specifications

Sample Type
DNA (Genomic), DNA/RNA (Pathogen), Total Nucleic Acids, DNA/RNA (Viral)
For Use With (Application)
Real-Time PCR, Sequencing
High-throughput Compatibility
High-throughput Compatible
Product Line
MagMAX™
Quantity
100 tests
Shipping Condition
Room Temperature
Target
Bacteria, Blood, DNA, Liquid Samples (e.g. Serum), Other Samples, RNA, Tissue, Viral Samples
Unit Size
100 tests

Contents & Storage

• MagMAX CORE Clarifying Solution, 45 mL, store at room temperature
• MagMAX CORE Bead Beating Tubes, 100 tubes (2 mL), store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text