Thermo Scientific™

Pierce™ Protease Inhibitor Mini Tablets, EDTA-free

Catalog number: A32955
Have Questions?
Thermo Scientific™

Pierce™ Protease Inhibitor Mini Tablets, EDTA-free

Catalog number: A32955
Have Questions?
Catalog Number
A32955
Unit Size
30 tablets
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A3295530 tablets
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Protease Inhibitor Mini Tablets, EDTA-free, contain broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation during cell lysis and protein extraction experiments.

Features of EDTA-free Protease Inhibitor Mini Tablets:
Improved formulation—tablets dissolve quickly to form a clear solution
Economical—more cost effective than other commercially available formulations for equivalent volumes
Convenient—ready-to-use, fully disclosed, broad-spectrum formulations
Compatible—use directly with Pierce BCA assays
EDTA-free—formulated without EDTA, a metalloproteinase inhibitor (only for EDTA-free formulations)

Pierce Protease Inhibitor Mini Tablets protect proteins from proteolytic degradation through inhibition of the serine proteases, cysteine proteases, aspartic acid proteases, and aminopeptidases that are typically present in cell lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin, and pepstatin A. Stable for storage at 4°C for up to 12 months, each mini tablet is sufficient for 10 mL of solution, and is suitable for reagent preparation prior to protein extraction from tissue and cultured cells.

Proteases make up a large category of enzymes, including endopeptidases and exopeptidases, which catalyze the hydrolytic breakdown of proteins into peptides or amino acids. Proteases reside in cellular compartments, but upon cell lysis the proteases are released, resulting in the degradation of valuable proteins. For this reason, it is common practice to add multiple compounds known to inactivate or inhibit proteases to cell lysis buffers or cell extracts.

To use Pierce Protease Inhibitor Mini Tablets, simply dissolve 1 tablet in 10 mL of buffer or lysate. The protease inhibitor formulation is compatible with most detergent-based cell lysis reagents, including Pierce Cell Lysis solutions. Pierce Protease Inhibitor Mini Tablets do not contain phosphatase inhibitors. If necessary, phosphatase inhibitors, such as Pierce Phosphatase Inhibitor Mini Tablets, can be added along with the protease inhibitor tablets.

Related products
Pierce Protease Inhibitor Mini Tablets
Pierce Protease Inhibitor Tablets
Pierce Phosphatase Inhibitor Mini Tablets
Pierce Protease and Phosphatase Inhibitor Mini Tablets

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
EDTA-Free
Product Type
Protease Inhibitor
Shipping Condition
Room Temperature
Form
Solid
Product Line
Pierce™
Sufficient For
10 mL Solution
Inhibitors
Protease Inhibitor
Quantity
30 Tablets

Contents & Storage

1 Vial, Store at 2°C to 8°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text