Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Fe-NTA Phosphopeptide Enrichment Kit |
For Use With (Equipment) | Microcentrifuge |
Product Type | Phosphopeptide Enrichment Kit |
Product Line | High Select™ |
Label or Dye | Unlabeled |
Shipping Condition | Wet Ice |
Starting Material | Protease-digested Protein |
Final Product Type | Peptides |
Detection Method | Mass Spectrometry |
Format | Kit |
Workflow Step | Peptide Enrichment |
Quantity | 30 Reactions |
Unit Size | 30 reactions |
Description | TiO2 Phosphopeptide Enrichment Kit |
For Use With (Equipment) | Microcentrifuge |
Product Type | Phosphopeptide Enrichment Kit |
Product Line | High Select™ |
Label or Dye | Unlabeled |
Shipping Condition | Wet Ice |
Starting Material | Protease-digested Protein |
Final Product Type | Peptides |
Detection Method | Mass Spectrometry |
Format | Kit |
Workflow Step | Peptide Enrichment |
Quantity | 24 Reactions |
Unit Size | 24 reactions |
Description | Graphite Spin Columns |
For Use With (Equipment) | Microcentrifuge, Mass Spectrometer |
Product Type | Column |
Product Line | Pierce™ |
Starting Material | Peptides, Protease-digested Protein |
Final Product Type | Peptides |
Detection Method | Mass Spectrometry |
Format | Spin Column |
Workflow Step | Sample Clean Up |
Quantity | 30 Columns |
Unit Size | 30 columns |
Description | TiO2 Phosphopeptide Enrichment Tips |
For Use With (Equipment) | Microcentrifuge, Mass Spectrometer |
Product Type | Phosphopeptide Enrichment Spin Tip |
Product Line | Pierce™ |
Starting Material | Peptides, Protease-digested Protein |
Final Product Type | Peptides |
Detection Method | Mass Spectrometry |
Format | Spin Column |
Workflow Step | Sample Enrichment |
Quantity | 96 Tips |
Unit Size | 96 tips |
Catalog Number | Specifications | Unit Size | Product Line | Description | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A32992 | 30 reactions | High Select™ | Fe-NTA Phosphopeptide Enrichment Kit | 457.65 Online price 477.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A32993 | 24 reactions | High Select™ | TiO2 Phosphopeptide Enrichment Kit | 632.65 Online price 662.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88302 | 30 columns | Pierce™ | Graphite Spin Columns | 176.65 Online price 184.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88303 | 96 tips | Pierce™ | TiO2 Phosphopeptide Enrichment Tips | 1,559.65 Online price 1,624.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|