Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
A33044 | 1 Adaptor |
This semi-skirted adaptor is for use with the 96-well Automated Thermal Cycler (ATC). It ensures uniform pressure is applied by the heated lid onto the plate, and also ensures that the plate is ejected smoothly without sticking. This adaptor is only for use with 96-well semi-skirted plates.
Share catalog number, name or link