PureLink™ Expi Endotoxin-Free Buffer Set
Inquire about OEM or Commercial Supply version of this product here.
PureLink™ Expi Endotoxin-Free Buffer Set
Invitrogen™

PureLink™ Expi Endotoxin-Free Buffer Set

The PureLink Expi Endotoxin-Free Buffer Set provides the same buffers included in the PureLink Expi Endotoxin-Free plasmid purification kits, madeRead more
Have Questions?
Catalog number A33074
Price (USD)/ 1 set
619.65
Online exclusive
650.00 
Save 30.35 (5%)
-
Add to cart
Price (USD)/ 1 set
619.65
Online exclusive
650.00 
Save 30.35 (5%)
Add to cart
PureLink™ Expi Endotoxin-Free Buffer Set
Catalog numberA33074
Price (USD)/ 1 set
619.65
Online exclusive
650.00 
Save 30.35 (5%)
-
Add to cart
The PureLink Expi Endotoxin-Free Buffer Set provides the same buffers included in the PureLink Expi Endotoxin-Free plasmid purification kits, made available separately here for those applications that would benefit from additional buffers. The Buffer Set includes all buffers and endotoxin-free components needed to enable 30 Maxi, 4 Mega, or 2 Giga PureLink Expi Endotoxin-Free plasmid purifications. Specifically, the buffer set includes RNAse A, resuspension buffer (R3), lysis buffer (L7), precipitation buffer (N3), equilibration buffer (EQ1), wash buffer (W8), elution buffer (E4), TE buffer (TE), endotoxin removal buffer, lysis indicator, and endotoxin-free water.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Sample TypeBacteria
ScaleMaxi
Expression SystemMammalian
For Use With (Application)Cloning, PCR
High-throughput CompatibilityManual Protocols
Prep Scale> 1 mg (Large-Scale) Plasmid DNA, > 200 μg (Large-Scale) Plasmid DNA
Product LinePureLink™
Product TypeBuffer Set
Quantity1 Set
Shipping ConditionRoom Temperature
TargetPlasmid DNA
Test Time90 min.
Unit Size1 set
Contents & Storage
• Resuspension buffer
• Lysis indicator
• RNase A
• Lysis buffer
• Precipitation buffer
• Endotoxin removal buffer
• Wash buffer
• Elution buffer
• TE buffer