CultureOne™ Supplement (100X)
Culturing neurons? Try our new Gibco™ Poly-D-Lysine Solution.
Increase cell survival by more than 50% with new Gibco™ B-27™ Plus Neuronal Culture System »
CultureOne™ Supplement (100X)
Gibco™
CultureOne™ Supplement (100X)
CultureOne Supplement is a chemically-defined, serum-free supplement designed to significantly improve the differentiation of neural stem cells (NSCs) to neurons.Read more
Have Questions?
Catalog NumberQuantity
A33202015 mL
Catalog number A3320201
Price (USD)
246.00
Each
Add to cart
Quantity:
5 mL
Price (USD)
246.00
Each
Add to cart
CultureOne Supplement is a chemically-defined, serum-free supplement designed to significantly improve the differentiation of neural stem cells (NSCs) to neurons. Compared to conventional differentiation methods where NSCs can overgrow and become burdensome, CultureOne Supplement eliminates more than 75% of contaminating neural progenitor cells with minimal cell death and no effect on other kinase-mediated pathways. The resulting superior neuronal cell cultures of evenly distributed, differentiated neurons enable improved downstream assays, accelerated neuronal maturation, and seamless maintenance for five weeks or more.
Each vial of CultureOne Supplement contains a 100X 5-mL liquid reagent that can be added easily to any conventional neuronal differentiation medium, such as Neurobasal Medium with B-27 Supplements. After two weeks of differentiation with CultureOne Supplement, the supplement can be removed and the resulting cultures will contain considerably fewer progenitor cells for five weeks or more.
Improved downstream assays
Analysis of NSC-derived neurons can be difficult due to the presence of overgrown progenitor cells in the culture. These invasive cells can make imaging and electrophysiology experiments challenging and call into question the physiological relevance of gene expression assays. CultureOne Supplement reduces proliferating progenitor cells, giving you a cleaner and more relevant neuronal cell population, which significantly improves the assay experience and outcome.
Accelerated maturation of differentiated neurons
With CultureOne Supplement, NSC-derived neurons express mature physiological markers faster and develop longer neurites than untreated NSC-derived neuronal cultures. CultureOne Supplement-treated neurons also express significantly higher numbers of voltage gated calcium ion channels, which are an important marker for neural maturity and excitability.
Longer culture of NSC-derived neurons
For experiments that require NSC-derived neurons to mature for long periods of time, CultureOne Supplement seamlessly enables maintenance of enriched, differentiated neurons for five weeks or longer. By contrast, conventional differentiation methods result in proliferating neural progenitor cells, which tend to clump and cause the culture to peel off of the plate.
For established protocols using CultureOne Supplement, access our Gibco Neurobiology Protocol Handbook.
View publications using Gibco neurobiology products at our Gibco CellCite.
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
DescriptionNeural Cells, Serum-Free
Product LineCultureOne™
Quantity5 mL
Shipping ConditionDry Ice
Tested ForEndotoxin, Sterility
FormLiquid
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -20°C) and protect from light.
Frequently asked questions (FAQs)
What are the storage conditions for CultureOne Supplement? Can I store it at 4 degrees C and for how long? Can I aliquot unused product and freeze it at -20 degrees C?
We recommend that you thaw the CultureOne Supplement and then aliquot it out for working stocks and re-freeze the aliquots. We have found that the product can undergo up to 3 freeze-thaws without loss of activity. CultureOne-supplemented media is good for 2 weeks stored at 4 degrees C in the dark.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
I am using the B-27 Plus Neuronal Culture System. What do I do if my neuronal cells are not attaching or are clumped?
In primary neuronal cultures, this is typically a function of uneven coating or if neurons are plated at too high of a density. In NSC differentiation experiments, this could be due to overproliferation of neural progenitor cells. Differentiating with the B-27 Plus Neuronal Culture System and CultureOne Supplement can eliminate more than 75% of neural progenitor cells and the clumps they form. Please see the CultureOne Supplement User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0016204_CultureOneSupplement_UG.pdf) for more information.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
I am planning to use the B-27 Plus Neuronal Culture System. How can I control the number of glial cells in my neuronal culture?
Add CultureOne Supplement at day 0 with the B-27 Plus Neuronal Culture System to fully suppress both astrocytes and oligodendrocytes with no detrimental effect on neurons. Delaying the addition of CultureOne Supplement to later time points results in increasing levels of astrocytes. Refer to the CultureOne Supplement User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0016204_CultureOneSupplement_UG.pdf) for more information.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What specific types of neurons will CultureOne Supplement work with?
CultureOne Supplement accelerates the maturation of all types of differentiating neurons.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I thaw CultureOne Supplement and re-freeze it?
CultureOne Supplement can be frozen and thawed up to 3 times without affecting the product performance.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link