Applied Biosystems™

SeqStudio™ Cathode Buffer Container

Catalog number: A33401
Have Questions?
Applied Biosystems™

SeqStudio™ Cathode Buffer Container

Catalog number: A33401
Have Questions?
Catalog Number
A33401
Unit Size
4 containers
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A334014 containers
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems SeqStudio Cathode Buffer Container (CBC) contains running buffer to support Sanger sequencing and fragment analysis applications on the SeqStudio Genetic Analyzer System. The container has two separate compartments: the left compartment provides the cathode buffer for electrophoresis and the right compartment provides capillary wash and spent polymer waste ejection functionality between injections. Store the Cathode Buffer Container at 2–8°C before use.

Key features:
• Ready-to-use, disposable container with running buffer
• Buffer usage tracking with radio frequency identification (RFID)
• Two week on-instrument shelf life after installation
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line
SeqStudio
Type
Cathode Buffer Container
Shipping Condition
Approved for shipment on Wet or Dry Ice

Contents & Storage

Store in refrigerator (2–8°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text