Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

Lysis/Binding Buffer for Dynabeads™ mRNA Purification Kits

Catalog number: A33562
Have Questions?
Invitrogen™

Lysis/Binding Buffer for Dynabeads™ mRNA Purification Kits

Catalog number: A33562
Have Questions?
Catalog Number
A33562
Unit Size
60 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A3356260 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This Lysis/Binding Buffer is included in the following Dynabeads™ mRNA purification kits: Cat. Nos. 61011, 61012, and 61021. It is made available separately for applications that require more Lysis/Binding Buffer than is provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line
Dynabeads™
Quantity
1 x 60 mL
Shipping Condition
Room Temperature

Contents & Storage

1 x 60 mL, store (at 2°C to 8°C)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text