PI(5)P Lipid Kinase Substrate
PI(5)P Lipid Kinase Substrate
Invitrogen™
PI(5)P Lipid Kinase Substrate
The PI(5)P Lipid Kinase Substrate has been specifically optimized for use with Type II PIP kinases (e.g., PIP4K2A). The substrateRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A335965000 nmol
A33593400 nmol
Catalog number A33596
Price (USD)/ Each
8,020.00
-
Add to cart
Quantity:
5000 nmol
Price (USD)/ Each
8,020.00
Add to cart
PI(5)P Lipid Kinase Substrate
Catalog numberA33596
Price (USD)/ Each
8,020.00
-
Add to cart
The PI(5)P Lipid Kinase Substrate has been specifically optimized for use with Type II PIP kinases (e.g., PIP4K2A). The substrate consists of solubilized phosphatidylinositol 5-phosphate (PI(5)P) in an aqueous buffer, and is used in combination with our Adapta™ Universal Kinase Assay to help identify unique kinase inhibitors.
• Validated for use with Thermo Fisher Scientific lipid kinases
• Available in multiple pack sizes for convenient use
• Also available in our SelectScreen™ Kinase Profiling Service
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormPowder
Product LineInvitrogen™
Quantity5000 nmol
SubstratePhosphatidylinositol 5-phosphate (PI(5)P)
Substrate TypeKinase Substrate
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Phosphatidylinositol 5-phosphate (PI(5)P) in 30 mM HEPES pH 7.5 buffer.