Thermo Scientific™

NeuCode™ Lysine-080

Catalog number: A33613
Have Questions?
Thermo Scientific™

NeuCode™ Lysine-080

Catalog number: A33613
Have Questions?
Catalog Number
A33613
Unit Size
50 mg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A3361350 mg
-
A33614500 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific™ NeuCode™ amino acids augment the level of multiplexing achievable for the metabolic labeling of proteins for mass spectrometry analysis. NeuCode™ Lysine-080 (3,3,4,4,5,5,6,6-D8 L-Lysine-2HCl) may also be used with traditional SILAC to improve flexibility of multiplexing options or to reduce complexity of analysis.

General features of NeuCode SILAC labeling:
• Labeling efficiency—100% label incorporation into proteins of living cells without toxicity
• Compatible—may be multiplexed with existing SILAC amino acids
• Time-saving—not necessary to label to 100% incorporation if only using heavy amino acids
• High-quality supplements—heavy amino acids with >98% isotope purity

Stable isotope labeling with amino acids in cell culture (SILAC) is a powerful method to identify and quantify relative differential changes in complex protein samples. NeuCode metabolic labeling is similar to SILAC, but differs in that the labeling only utilizes heavy amino acids. The increased multiplexing capability of NeuCode amino acids is possible through the use of mass defects from extra neutrons in the stable isotopes. These small mass differences may be resolved on high resolution mass spectrometers (Thermo Scientific™ Orbitrap™ Elite™, Q Exactive™, Orbitrap™ Fusion™ Tribrid™, and Orbitrap™ Fusion™ Lumos™ Tribrid™ mass spectrometers). Use of only heavy amino acids eliminates the need for 100% incorporation of amino acids used for SILAC (both heavy and light), and may be especially useful for studies with primary cells.

NeuCode amino acids are used together with specialized cell culture media that are deficient in essential amino acids. Heavy L-lysine is used for SILAC analysis of peptides that have been digested with trypsin or LysC. NeuCode Lysine-080 may be used with 4,4,5,5-D4 L-lysine or 13C6 15N2 L-lysine for duplex experiments and may be combined with light lysine for three-plex experiments.

Related products
L-Lysine-2HCl, 4,4,5,5-D4 for SILAC
L-Lysine-2HCl, 13C6, 15N2 for SILAC
Fetal Bovine Serum, dialyzed, US origin
DMEM Media for SILAC

NeuCode is a trademark of WARF.

Specifications

Label Type
Mass Isotopes of Amino Acids
Form
Solid
Final Product Type
Molecule
Quantity
50 mg
Product Type
Lysine

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text