Informational:Inquire about OEM or Commercial Supply version of this product here.
Applied Biosystems™

MagMAX™ Plant RNA Isolation Kit

Catalog number: A33899
Have Questions?
Applied Biosystems™

MagMAX™ Plant RNA Isolation Kit

Catalog number: A33899
Have Questions?
Catalog Number
A33899
Unit Size
384 preps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
96 preps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1000 preps
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A33899384 preps
-
A3378496 preps
-
A471571000 preps
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The MagMAX Plant RNA Isolation Kit is designed for automated high-throughput or manual purification of total RNA from wide variety of plant species and tissue types. It utilizes paramagnetic bead technology, enabling high yields and robust performance. The high binding capacity, uniform particle size, and rapid magnetic response of MagMAX magnetic beads makes the technology ideal for high-throughput automatic nucleic acid purification, as well as for manual nucleic acid purification by low- throughput users. The resulting high quality purified RNA is free of proteins, nucleases, and other contaminants or inhibitors, and can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, and other enzymatic reactions.

Versatile—suitable for any plant type and part, including woody, polyphenol-rich, and lignified samples
High yields of pure total RNA
High quality—isolates intact total RNA, free of inhibitors and gDNA contamination. Isolated RNA is suitable for long term storage at -20°C.
Flexible—easily adjustable for high-throughput automation platforms and manual processing

Applications
The MagMAX Plant RNA Isolation Kit is designed for fast extraction of high purity RNA suitable for all conventional molecular biology applications, including:
• RT-PCR
• RT-qPCR
• Northern blotting
• Nuclease protection assay

Benefits of MagMAX magnetic bead–based purification
The magnetic bead–based purification format of the kit allows for easy scaling of the number of samples processed either manually with a magnetic stand or with a magnetic particle processor such as the KingFisher Duo Prime or KingFisher Flex processors.

Magnetic beads offer many benefits compared to other technologies for isolating nucleic acids. Beads bind the RNA and DNA more efficiently than glass fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of undigested tissue particulates clogging these items during the extraction process. This clogging issue is of particular concern with fibrous plant tissue samples that that are not fully lysed during the disruption and homogenization steps.

Optimized bead surface chemistry
The Dynabeads MyOne Carboxylic Acid utilized in the MagMAX Plant RNA Isolation Kit has an optimized surface chemistry with high specific surface area that allows efficient kinetics and high sensitivity in nucleic acid capture. These aspects make Dynabeads magnetic technology ideal for the recovery of the RNA targets found in a variety of plant samples. Once captured, the RNA is then eluted in 100 μL and ready for downstream applications.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type
Plant Samples
High-throughput Compatibility
High-throughput Compatible, Manual Protocols, Automated Protocols
For Use With (Application)
Sequencing, RT-PCR (Endpoint), Real-Time PCR
Product Line
MagMAX™
Quantity
384 Preps
Shipping Condition
Room Temperature
Target
Total RNA

Contents & Storage

• 60 mL lysis buffer, store at room temperature
• 4 vials Dnase I (lyophilized), store at -5 to -30°C after reconsitution
• 4 x 1 mL DNase I reconstitution buffer (to be added to DNase I vial)
• 12 x 1 mL manganese chloride solution, store at -5 to -30°C
• 8 x 1.4 mL RNA binding beads, store at 2°C to 8°C
• 4 x 12 mL DNase I buffer (2X), store at -20° ± 5°C
• 4 x 110 mL Wash Solution 1 concentrate, store at room temperature
• 4 x 50 mL Wash Solution 2 concentrate, store at room temperature
• 4 x 20 mL rebinding buffer, store at room temperature
• 4 x 30 mL elution buffer, store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text