Thermo Scientific™

SILAC Protein Quantitation Kit (LysC), RPMI 1640

Catalog number: A33971
Have Questions?
Thermo Scientific™

SILAC Protein Quantitation Kit (LysC), RPMI 1640

Catalog number: A33971
Have Questions?
Catalog Number
A33971
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A339711 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific SILAC Protein Quantitation Kit - RPMI 1640 is used for the specific analysis of protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC).

Features of the SILAC Protein Quantitation Kit - RPMI 1640:
• Efficient—up to 100% label incorporation into proteins of living cells
• Reproducible—minimizes intra-experimental variability caused by differential sample preparation
• Flexible—media deficient in both L-lysine and L-arginine, allowing for more complete proteome coverage through dual amino acid isotope labeling
• Compatible—label proteins expressed in a wide variety of mammalian cell lines adapted to grow in RPMI 1640, including HeLa, 293T, COS7, U2OS, A549, A431, HepG2, NIH 3T3, Jurkat and others
• High-quality supplements—heavy amino acids with >99% isotope purity; dialyzed FBS tested to help ensure that it is sterile, endotoxin-free, and cell culture compatible

Applications of SILAC Protein Quantitation Kit:
• Quantitative analysis of relative changes in protein abundance from different cell treatments
• Quantitative analysis of proteins for which there are no antibodies available
• Protein expression profiling to study normal vs. disease cells
• Identification and quantification of hundreds to thousands of proteins in a single experiment

SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of 'heavy' 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization, and quantitation of proteins.

Because peptides labeled with heavy and light amino acids are chemically identical, they co-elute during reverse-phase column pre-fractionation and therefore are detected simultaneously during MS analysis. The relative peak intensities of multiple isotopically distinct peptides from each protein are then used to determine the average change in protein abundance in the treated sample.

Each kit includes all necessary reagents to isotopically label cells with 13C6 L-lysine-2HCl, including media, heavy and light amino acids, and dialyzed serum. Heavy L-arginine-HCl derivatives (Cat. Nos. 88210, 89990) are available separately and can be combined with Pierce SILAC Protein Quantitation kits to enhance peptide isotope label coverage. When combined with Thermo Scientific Protein/Peptide Sample Enrichment Products, Pierce SILAC Protein Quantitation kits also enable MS analysis of low-abundance proteins such as cell surface proteins, organelle-specific proteins, and post-translational protein modifications such as phosphorylation or glycosylation.

Related products
Dialyzed Fetal Bovine Sera, US Origin
Pierce Trypsin Protease, MS Grade
RPMI 1640 Medium for SILAC
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
SILAC Protein Quantitation Kit
Label or Dye
Mass Isotope of Amino Acid
Shipping Condition
Dry Ice
Starting Material
Cell Lysate
Final Product Type
Protein
Format
Kit
Quantity
1 Kit

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text