Applied Biosystems™

AgriSeq™ HTS Library Kit

Catalog number: A34144
Have Questions?
Applied Biosystems™

AgriSeq™ HTS Library Kit

Catalog number: A34144
Have Questions?
Catalog Number
A34144
Unit Size
960 reactions
Price (USD)
Quantity
Catalog Number
Unit Size
9600 reactions
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
A34144960 reactions
A341439600 reactions
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems AgriSeq HTS Library Kit is specifically designed for high-throughput preparation of amplicon libraries for targeted genotyping-by-sequencing (GBS) applications in agrigenomics.

Key product features include:
• Exceptional uniformity of multiplexed target amplification generates robust, reproducible results
• Simplified workflow enables high-throughput sample processing
• Convenient configuration contains library normalization reagents and volumes sufficient for automated platforms

Excellent data quality
AgriSeq library preparation has been optimized for targeted GBS of panels ranging from 100–5000 amplicons. With a highly efficient and specific multiplex technology, AgriSeq amplification can generate libraries with high uniformity (87.2–99.3%), percent on-target reads (85.7–99.7%), and overall genotype call rates (87.5–99.7%) based on a set of nine recently designed panels ranging in size from 62–3000 amplicons across seven different species. The figure below shows an average of 98.2% uniformity for a 3000-amplicon panel across 192 samples (i.e., 98% of bases are covered by at least 0.2X of the average read depth). Even coverage is significant in that it reduces the amount of sequencing required to achieve consistent genotype calls, thus reducing the sequencing cost per sample. In comparison, weaker multiplexing performance is reported with the competitor technology TruSeq, which provides only 88% average uniformity and a reported 1536 maximum multiplexing capability (see reference).

High throughput sample processing
The AgriSeq Library workflow (see figure below) can be performed in either 96- or 384-well plates for high-throughput sample processing using as little as 10 ng of DNA input per reaction. The workflow has been optimized to minimize sample handling and ancillary reagent usage through the processing of pooled samples. This allows the workflow to be completed with minimal pipetting steps, helping to reduce the potential for error, as well as simplifying automation setup on common third party platforms such as Tecan or Hamilton systems. Standard data analysis processing includes automated quality assessment, alignment, and genotype calling using the included Torrent Suite software and Variant Caller plug-in without the need for a bioinformatician, expensive data storage, or third party programs.

Conveniently packaged containing library normalization reagents & volumes for automation
The AgriSeq HTS Kit is offered in large pack sizes (960 or 9600 reactions) to accommodate high throughput agrigenomics projects while minimizing the cost and waste of unnecessary packaging. The AgriSeq HTS Kit includes not only the reagents for library preparation, but all of the components required for library normalization to help achieve consistent results between samples, and sufficient overage has been provided to accommodate automated workflows. With a low price per sample exclusively available for agricultural customers, AgriSeq targeted GBS is an economical way to generate high-throughput genotyping data to advance your breeding programs.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Kit
Sample Type
DNA
Sequencing Type
Genome and DNA Sequencing
For Use With (Equipment)
Ion GeneStudio S5 System
For Use With (Application)
Sequencing
Libraries
Targeted Sequencing Library
No. of Reactions
960 Reactions
Product Line
AgriSeq™
Product Type
HTS Library Kit
Shipping Condition
Approved for shipment at Room Temperature or on Wet or Dry Ice
Workflow Step
Library Generation

Contents & Storage

Store in freezer (-5 to -30°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text