Ion Torrent™

Ion 550™ Chip Kit

Catalog number: A34538
Have Questions?
Ion Torrent™

Ion 550™ Chip Kit

Catalog number: A34538
Have Questions?
Catalog Number
A34538
Unit Size
8 chips
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 chips
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A345388 chips
-
A345374 chips
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ion 550 Chip Kit contains four (Cat. No. A34537) or eight (Cat. No. A34538) barcoded chips for sample tracking and sequencing with the Ion GeneStudio S5 Plus, the Ion GeneStudio S5 Prime, and Ion S5 XL sequencing systems. The Ion 550 Chip generates 100–130 million sequencing reads by electronically detecting polymerase-driven base incorporation without the use of fluorescence. By eliminating the use of an optical detection system, this advance in next-generation sequencing technology allows for rapid sequencing times of as little as 2.5 hours for 200 bp sequencing.

The Ion 550 Chip Kit offers:
• Support for a variety of research applications, including sequencing of up to four human exomes, cell-free nucleic acid applications, transcriptome profiling, and targeted resequencing of gene panels
• A rapid run-time of as little as 2.5 hours for 200 bp sequencing
• Sample tracking functionality from an integrated barcode
Notes:The Ion 550 Chip is only compatible with the Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, and Ion S5 XL systems.
Use this chip with the Ion 550 Kit—Chef for automated template preparation and chip loading.

Specifications

Product Line
Ion Torrent™
For Use With (Application)
Comparative Sequencing (Germline Mutations - 50:50)
For Use With (Equipment)
Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, Ion S5 XL Sequencing System
Green Features
Sustainable disposal
Product Type
Chip Kit
Quantity
8 Chips
Shipping Condition
Room Temperature
Unit Size
8 chips

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text