ReproSeq™ PGS Kit with Ion 530™ Chips (96 samples/run)
ReproSeq™ PGS Kit with Ion 530™ Chips (96 samples/run)
Ion Torrent™

ReproSeq™ PGS Kit with Ion 530™ Chips (96 samples/run)

The ReproSeq PGS Kit for the Ion S5 System provides all the reagents and materials necessary for a rapid, cost-effective,Read more
Have Questions?
Catalog NumberQuantity
A34901384 samples
Catalog number A34901
Price (USD)
54,400.00
Each
-
Add to cart
Quantity:
384 samples
Recurring order eligible. Learn more »
Price (USD)
54,400.00
Each
Add to cart
Ask our AI about this Product
The ReproSeq PGS Kit for the Ion S5 System provides all the reagents and materials necessary for a rapid, cost-effective, and comprehensive next-generation sequencing (NGS) workflow for aneuploidy analysis. When used with an Ion S5 sequencing system and Ion Chef System for automated template preparation, the kit enables a simple, end-to-end workflow for the detection of aneuploidies across all 24 chromosomes (22 autosomes and the X and Y chromosomes) for research labs interested in reliably assessing pre-implantation embryo biopsy samples.

Using as little as 6 pg of DNA from a single cell, or input from multiple cells, the ReproSeq PGS Kit for the Ion S5 System enables accurate detection of whole chromosome and chromosome arm copy number events (down to 17 Mbp copy number events*) in as little as 10 hours**. Three kit configurations provide comprehensive sample throughput flexibility enabling cost-effective screening of 16, 24, or 96 samples per run. When combined with analysis using Ion Reporter v5.4 or later, data interpretation is enhanced with new functionality such as mosaicism detection, gender masking, and improved data plotting.

Cost-effective NGS workflow with sample throughput flexibility
The ReproSeq PGS Kit for the Ion S5 System delivers scalability and flexibility through three sequencing chips—the Ion 510 Chip, Ion 520 Chip, and Ion 530 Chip—to support various levels of sample throughput. Choose the kit that fits your throughput needs to process 16, 24, and up to 96 samples to achieve throughput flexibility that scales up to improve cost efficiencies and allows your laboratory to respond to sample fluctuation and increases in sample volume.

Rapid turnaround for time-sensitive samples
Leveraging the speed of semiconductor sequencing and automated template preparation by isothermal amplification on the Ion Chef System, the ReproSeq PGS Kit for the Ion S5 System offers a fast benchtop NGS workflow for aneuploidy analysis studies. Go from a single cell or multiple cells isolated from a pre-implantation embryo to analyzed data typically in less than 10 hours when processing up to 16 samples, 10 hours for 24 samples, or 13 hours for up to 96 samples. This rapid turnaround time enables reliable identification of chromosomally normal embryos for time-sensitive genetic research.

The ReproSeq PGS Kit with Ion 530 Chips (96 samples/run) includes all reagents for processing a total of 384 samples through a workflow that includes whole genome amplification, library construction, template preparation and chip loading on the Ion Chef instrument, and sequencing using an Ion S5 System.

The kit contains:
• Ion SingleSeq 96 Kit (Cat. No. A34763)—includes reagents to extract, amplify, and barcode genomic DNA for preparing libraries from pre-implantation embryo biopsy samples, a process that can be completed in less than 5 hours (includes hands-on time)
• Ion S5 ExT Chef Supplies, Ion S5 Chef Solutions, and Ion S5 ExT Chef PGS Reagents—contains pre-packaged single-use cartridges for automated template preparation and chip loading (5 hours, includes hands-on time) whereby library DNA is clonally amplified onto a bead surface through an emulsion-based isothermal amplification reaction
• Ion S5 ExT Sequencing Solutions, Ion S5 ExT Sequencing Reagents—simplifies sequencing with ready-to-use, disposable reagent cartridges for fast and simple instrument set-up, with less than 15 minutes hands-on time, for a sequencing chemistry supporting advanced data quality and accuracy
• Ion 530 Chip Kit (Cat. No. A27763)—contains 4 barcoded chips per kit for sample tracking and sequencing runs using the Ion S5 System

Notes
• This kit is only compatible with the Ion Chef System and Ion S5 systems. The Ion S5 XL System is recommended for rapid turnaround time between sequencing runs. Use of the kits with the Ion S5 System requires significantly more analysis time between runs.
• The ReproSeq PGS Kit is compatible with Torrent Suite Software 5.4 and later. Update your Torrent Server to the latest available version of Torrent Suite Software before using these kits. For data analysis, Ion Reporter Software 5.4 or later is recommended.

*Detecting copy number events <48 Mbp using the ReproSeq™ PGS Kit with Ion 530™ Chips (96 samples/run) requires running ≤24 samples on an Ion 530 Chip
**Sample to analyzed results turnaround time for the ReproSeq PGS Kit with Ion 510 Chips (16 samples/run) is 9.6 hours.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Shotgun Sequencing, Rapid Sequencing
For Use With (Equipment)Ion GeneStudio S5 System, Ion PGM™ System, Ion Chef™ System
No. of Samples384
Product LineIon 530™, ReproSeq™
Product TypePGS Kit
Quantity384 samples
Shipping ConditionApproved for shipment on Wet or Dry Ice
FormatKit
Unit SizeEach
Contents & Storage
• Ion SingleSeq 96 Kit, 4 boxes, –10 to –30°C
• Ion S5 ExT Chef Supplies, 4 boxes, 15–30°C
• Ion S5 Chef Solutions, 4 cartridges, 15–30°C
• Ion S5 ExT Chef PGS Reagents, 4 cartridges, –10 to –30°C
• Ion S5 ExT Sequencing Solutions, 1 box, 15–30°C
• Ion S5 ExT Sequencing Reagents, 4 cartridges, –10 to –30°C
• Ion 530 Chip Kit, 4 chips, 15–30°C
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

How many total samples can I process with the Ion ReproSeq PGS Kit with Ion 530 Chips?

384 samples in total (4 runs with 96 samples/run) can be processed with the Ion ReproSeq PGS Kit with Ion 530 Chips.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How many samples per run can I process with the Ion ReproSeq PGS Kit with Ion 530 Chips?

96 samples per run can be processed with the Ion ReproSeq PGS Kit with Ion 530 Chips.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What does the Ion ReproSeq PGS Kit with Ion 530 Chips (96 samples/run) include, and what are the storage conditions for each component?

*Ion SingleSeq 96 Kit, 4 boxes, -10 to -30 degrees C
*Ion S5 ExT Chef Supplies, 4 boxes, 15-30 degrees C
*Ion S5 Chef Solutions, 4 cartridges, 15-30 degrees C
*Ion S5 ExT Chef PGS Reagents, 4 cartridges, -10 to -30 degrees C
*Ion S5 ExT Sequencing Solutions, 1 box, 15-30 degrees C
*Ion S5 ExT Sequencing Reagents, 4 cartridges, -10 to -30 degrees C
*Ion 530 Chip Kit, 4 chips, 15-30 degrees C

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

Does the Ion SingleSeq 96 Kit include reagents for template preparation, sequencing, and chips?

No, the Ion SingleSeq 96 Kit only includes reagents to prepare the library. For a detailed list, please go here (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0016712_IonReproSeqPGS_S5_UG.pdf).

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What is included in the Ion SingleSeq 96 Kit?

The Ion SingleSeq 96 Kit includes reagents to extract, amplify, and barcode genomic DNA for preparation of libraries from up to 96 samples. For a detailed list, please go here (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0016712_IonReproSeqPGS_S5_UG.pdf).

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Safety Data Sheets

Limited Use Label Licenses (LULL)

'

Notice to Purchaser: Our products are to be used for research purposes only. They may not be used for any other purposes, including, but not limited to, use in drugs, in vitro diagnostics purposes, therapeutics, or in humans. Our products may not be transferred to third parties, resold, modified for resale, or used to manufacture commercial products or to provide a service to third parties without prior written approval of Takara Bio USA, Inc.

Your use of this product is also subject to compliance with the licensing requirements listed below and described on the product's web page at http://www.takarabio.com. It is your responsbility to review, understand and adhere to any restrictions imposed by these statements.

Ion SingleSeq Kit contains PicoPLEX ™ technology developed and manufactured by Takara Bio USA, Inc. PicoPLEX ™ technology is covered by applicable US patents and corresponding patents outside the US.

'

Share catalog number, name or link

1x1 image pixel for data collection