Informational:Interested in a GMP LV-MAX Transfection Kit? Click here »
Gibco™

LV-MAX™ Transfection Kit

Catalog number: A35348
Have Questions?
Gibco™

LV-MAX™ Transfection Kit

Catalog number: A35348
Have Questions?
Catalog Number
A35348
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A353481 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Gibco LV- MAX Transfection Kit is an essential component of the LV-MAX Lentiviral Production System. It is formulated for the transient transfection of Gibco Viral Production Cells (high-density suspension-adapted human embryonic kidney (HEK) 293-derived cells). The included high-efficiency, cationic, lipid-based transfection reagent, lentiviral production supplement, and LV enhancer are designed to work together to produce the highest possible level of lentiviral particles from the Viral Production Cells when cultured in LV-MAX Production Medium.

• Designed specifically for transfection of high-density suspension cells to maximize lentiviral production
• Enables sustained, high-level production of high-density transiently-transfected cultures, achieving titer of up to 1 x 108 TU/mL (unconcentrated LVVs-GFP)
• Reproducible and scalable transfection results, giving users greater confidence in their results
• Easy, robust culture and transfection protocols—fits into any transient production workflowsThe kit includes three key components required to optimize your lentiviral production. These are only available in the kit and are not sold separately.
• LV-MAX transfection reagent—a high-efficiency, cationic, lipid-based transfection reagent uniquely designed for co-transfecting multiple plasmids into high-density Viral Production Cell cultures with high transfection efficiency and low toxicity
• LV-MAX supplement—an optimized, chemically defined, serum-free, protein-free, animal origin-free formulation designed to suppress cell growth during transfection and increase lentiviral vector production without compromising the cell viability and interfering with the lentiviral production process
• LV-MAX enhancer—a proprietary, animal origin-free formulation designed to boost cell lentiviral vector production

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Line
Established Cell Lines
Classification
Animal Origin-Free, Chemically Defined, Protein-Free, Serum-Free
Cell Type
293-derived Cells
Sample Type
Plasmid DNA
Transfection Technique
Lipid-based Transfection
For Use With (Application)
Transfection
Product Line
LV-MAX™
Product Type
Transfection Kit
Quantity
1 L
Serum Compatible
Yes
Shipping Condition
Wet Ice

Contents & Storage

• 50 mL LV-MAX Supplement
• 6 mL LV-MAX Transfection Reagent
• 40 mL LV-MAX Enhancer

Sufficient for 1 L LV production 

Store at 2–8°C, protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text