Thermo Scientific™

Pierce™ BS(PEG)5, No-Weigh™ Format

Catalog number: A35396
Have Questions?
Thermo Scientific™

Pierce™ BS(PEG)5, No-Weigh™ Format

Catalog number: A35396
Have Questions?
Catalog Number
A35396
Unit Size
10 x 1 mg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A3539610 x 1 mg
-
21581100 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
BS(PEG)5 is a bis-succinimide ester-activated PEG compound used for crosslinking between primary amines (—NH2) in proteins and other molecules. The N-hydroxysuccinimide ester (NHS) groups at either end of the PEG5 spacer react specifically and efficiently with lysine and N-terminal amino groups at pH 7-9 to form stable amide bonds.

Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging prevents the loss of reagent reactivity and contamination over time by eliminating the repetitive opening and closing of the vial. The format enables use of a fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.

Features of BS(PEG)5:

Reactive groups: sulfo-NHS ester (both ends)
Reactive towards: amino groups (primary amines)
• NHS esters react with—NH2 groups at pH 7-9, forming stable amide bonds
• Water-soluble; compare to non-pegylated BS3 (also soluble) and DSS (water-insoluble)
• Membrane-impermeable, allowing for cell surface labeling
• Irreversibly crosslink proteins or peptides by flexible PEG spacer arms
• Polyethylene glycol spacer arms help maintain conjugate solubility
• Pure compound with defined structure and molecular weight, ensuring reproducible protein-modification effects
• PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
• Ideal for small molecule or peptide conjugations

Alternative Names for BS(PEG)n:

• Bis-NHS-dPEG
• NHS-PEG-NHS
• BS(PEO)5
• Bis(NHS)PEO5
• BS-PEO9
• dPEG NHS ester
• Bis-succinimide ester PEG 400

Product References
Crosslinker Application Guide -- search for recent literature references for this product
Bioconjugate Techniques, Academic Press (Part No. 20036)

Related Products
BS(PEG)9 (PEGylated bis(sulfosuccinimidyl)suberate)
BS3 (bis(sulfosuccinimidyl)suberate)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Amine-Amine
Cleavable
No
Description
BS(PEG)5
Molecular Weight (g/mol)
532.5
PEGylated
Yes
Spacer Arm Length
21.7 Å
Cell Permeability
No
Shipping Condition
Wet Ice
Product Line
No-Weigh™, Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Homobifunctional
Reactive Moiety
Sulfo-NHS Ester
Spacer
Medium (10 to 30 Å)
Form
Powder
Quantity
10 x 1 mg
Solubility
Water
Format
Standard, Single-use
Water Soluble
Yes
Product Type
Crosslinker

Contents & Storage

Upon receipt store desiccated at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text