Acridine Orange (10 mg/mL in water)
Acridine Orange (10 mg/mL in water)
Invitrogen™

Acridine Orange (10 mg/mL in water)

Acridine Orange is a cell-permeant nucleic acid binding dye that emits green fluorescence when bound to dsDNA and red fluorescenceRead more
Have Questions?
Catalog NumberQuantity
A356810 mL
Catalog number A3568
Price (USD)
121.00
Each
-
Add to cart
Quantity:
10 mL
Price (USD)
121.00
Each
Add to cart
Ask our AI about this Product
Acridine Orange is a cell-permeant nucleic acid binding dye that emits green fluorescence when bound to dsDNA and red fluorescence when bound to ssDNA or RNA. This unique characteristic makes acridine orange useful for cell-cycle studies. Acridine orange has also been used as a lysosomal dye.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorOrange
Detection MethodFluorescence
Dye TypeCell-Permeant
Excitation Wavelength Range460/650 (RNA)
For Use With (Equipment)Fluorescence Microscope
Quantity10 mL
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypeDye
SubCellular LocalizationNucleic Acids, Nucleus
Unit SizeEach
Contents & Storage
Store in refrigerator 2°C to 8°C and protect from light.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link