Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

PureLink™ Fast Low-Endotoxin Maxi Plasmid Purification Kit

Catalog number: A35895
Have Questions?
Invitrogen™

PureLink™ Fast Low-Endotoxin Maxi Plasmid Purification Kit

Catalog number: A35895
Have Questions?
Catalog Number
A35895
Unit Size
10 preps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 preps
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A3589510 preps
-
A3622820 preps
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The PureLink Fast Low-Endotoxin Maxi Plasmid Purification Kit uses next-generation columns with advanced silica membranes that enable isolation of transfection grade (less than 1 EU/μg) plasmid DNA in about 30 minutes. The simple precipitation-free protocol yields up to 1.5 mg plasmid, suitable for standard transfections and all molecular biology applications such as cloning and sequencing.

NOTE: The use of a vacuum manifold is required to perform this procedure. We recommend our FastVac Vacuum Manifold.

Advantages of the PureLink Fast Low-Endotoxin Maxi Plasmid Purification Kit include:
Ultra-fast protocol—innovative silica-based columns enable purification of transfection grade plasmid DNA in under 30 minutes
Low endotoxin plasmid—the resulting plasmid contains less than 1 EU/μg endotoxins
High yields—isolate up to 1.5 mg of high-quality plasmid DNA
Colored buffers—the kit features colored buffers that permit error-free visualization of complete bacterial cell lysis and subsequent neutralization
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Kit
Isolation Technology
Silica Column
Scale
Maxi
For Use With (Application)
Cloning, PCR
No. of Reactions
10 Preps
Prep Scale
Maxi input culture of 100-500 mL to obtain a yield of up to 850 μg
Product Line
PureLink™
Product Type
Plasmid Purification Kit
Quantity
10 Preps
Shipping Condition
Room Temperature
Test Time
30 min.

Contents & Storage

• Resuspension buffer, store at 4°C upon reciept
• Lysis buffer, store at room temperature
• Precipitation buffer, store at room temperature
• Binding buffer, store at room temperature
• Wash buffer 1, store at room temperature
• Wash buffer 2 (concentrate), store at room temperature
• Elution buffer, store at room temperature
• Column assemblies, store at room temperature
• Syringe filters, store at room temperature
• Syringe plungers, store at room temperature
• Collection tubes, store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text