1x1 image pixel for data collection
FastVac™ Vacuum Manifold
FastVac™ Vacuum Manifold
Thermo Scientific™
FastVac™ Vacuum Manifold
The Thermo Scientific FastVac Vacuum Manifold features durable chemical-resistant construction and is capable of processing up to 20 samples simultaneouslyRead more
Have Questions?
Catalog NumberQuantity
A358991 Ea.
Catalog number A35899
Price (USD)
364.65
Online Exclusive
379.00
Save 14.35 (4%)
Each
In stock
Add to cart
Quantity:
1 Ea.
Price (USD)
364.65
Online Exclusive
379.00
Save 14.35 (4%)
Each
Add to cart
The Thermo Scientific FastVac Vacuum Manifold features durable chemical-resistant construction and is capable of processing up to 20 samples simultaneously using a vacuum, eliminating the need for multiple centrifugation steps and disposal of flow-through from collection tubes. The one-way Luer-Lock stopcocks are compatible with most nucleic acid purification columns and individually controlled, allowing the processing of as few as one sample at a time. Furthermore, the extra-large waste reservoir makes the FastVac Vacuum Manifold convenient for protocols that require the processing of large volumes, such as during plasmid DNA isolation using a PureLink Fast Low-Endotoxin plasmid purification kit.
Product features:
Capacity—capable of processing 1 to 20 samples simultaneously using columns and a vacuum source
Composition—consists of a linear, low-density polyethylene manifold body, one-way Luer-Lock stopcocks made of a polycarbonate body and polypropylene handle, a polypropylene vacuum hose connector, and a neoprene stopper
Compatibility—compatible with most nucleic acid purification kits (refer to the kit instruction manual for information regarding compatibility and processing with a vacuum manifold)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineFastVac™
Quantity1 Ea.
Shipping ConditionRoom Temperature
Product TypeVacuum Manifold
Unit SizeEach
Contents & Storage
• 1 vacuum manifold
• 20 Luer-Lock connectors
• 1 Neoprene stopper
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link