Ion Torrent™

Ion AmpliSeq™ Library Kit Plus

Catalog number: A35907
Have Questions?
Ion Torrent™

Ion AmpliSeq™ Library Kit Plus

Catalog number: A35907
Have Questions?
Catalog Number
A35907
Unit Size
96 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
24 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
384 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A3590796 reactions
-
448899024 reactions
-
A38875384 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The Ion AmpliSeq Library Kit Plus is designed for rapid preparation of amplicon libraries using Ion AmpliSeq panels. Ion AmpliSeq technology enables scalable, multiplex polymerase chain reactions (PCR) from 12- to 24,000-plex in a single well. Next-generation sequencing (NGS) libraries can be prepared using as little as 1 ng of starting material.

Features of the Ion AmpliSeq Library Kit Plus include:

  • Scalable and efficient library preparation
  • Easy and robust protocol
  • Barcoded sample multiplexing

Scalable and efficient library preparation

Using as little as 1 ng of starting material, highly multiplexed PCR amplification of thousands of genomic target regions yields superior coverage uniformity and specificity, without the need of a specialized microfluidics PCR platform. The primers contain proprietary modifications that enable removal of primer sequences during library preparation, thereby resulting in efficient target assessment during sequencing.

Easy and robust protocol

The Ion AmpliSeq Library Kit Plus uses a plate-based format for easier sample handling and tracking, and for compatibility with automation and high-throughput laboratories. The protocol is simple to use and enables high yield and uniformity, reliably and reproducibly. The resulting libraries are ready for downstream template preparation using the automated Ion Chef System (which also provides the option of automated library preparation using the Ion AmpliSeq Kit for Chef DL8 (Cat. No. A29024)) or the Ion OneTouch 2 System followed by sequencing. The intuitive Torrent Suite Software and optional Ion Reporter Software enables automation of data analysis.

Barcoded sample multiplexing

The Ion AmpliSeq Library Kit Plus includes reagents for generating amplicons using Ion AmpliSeq primers and preparing libraries from the resulting amplicons. The kit enables the preparation of barcoded libraries using Ion Xpress Barcode Adapters 1-96 kits or IonCode Barcode Adapters 1-384 kits. Barcoded libraries can be combined and loaded onto a single Ion chip to minimize the sequencing run time and cost, and to allow for accurate sample-to-sample comparisons.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Kit
Sample Type
DNA
Sequencing Type
Genome and DNA Sequencing
For Use With (Equipment)
Ion GeneStudio™ S5 Series Sequencer, Ion Chef™ System
For Use With (Application)
Sequencing
Libraries
Targeted Sequencing Library
No. of Reactions
96 Reactions
Product Line
Ion AmpliSeq™
Product Type
Library Kit Plus
Shipping Condition
Approved for shipment on Wet or Dry Ice
Workflow Step
Library Generation

Contents & Storage

• 6 tubes, store at -5°C to -30°C.
• 1 tube Low TE Buffer, store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text