The Attune NxT Blocker Bar Conversion Kit provides a 1.5-mm blocking bar for use on the Attune NxT Acoustic FocusingRead more
Have Questions?
Catalog number A35966
Price (USD)/ 1 kit
3,775.00
-
Add to cart
Price (USD)/ 1 kit
3,775.00
Add to cart
Attune™ NxT Blocker Bar Conversion Kit
Catalog numberA35966
Price (USD)/ 1 kit
3,775.00
-
Add to cart
The Attune NxT Blocker Bar Conversion Kit provides a 1.5-mm blocking bar for use on the Attune NxT Acoustic Focusing Cytometer, which is delivered with a 0.5 mm blocking bar. The standard 0.5-mm blocking bar accommodates cells or particles from 0.2–10 microns in size, while the larger blocking bar included in this kit is recommended for analysis of cells that are primarily larger than 10 microns. A larger blocking bar blocks more forward scatter light and allows the detection of larger cells. An experienced Thermo Fisher Scientific engineer can install the larger blocking bar on any Attune NxT cytometer, simplifying the instrument settings optimization.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineAttune™
Quantity1 kit
Shipping ConditionRoom Temperature
For Use With (Equipment)Attune NxT Acoustic Focusing Cytometer