Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Top14 Abundant Protein Depletion Mini Spin Columns |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Column |
Product Line | High Select™ |
Label or Dye | Unlabeled |
Shipping Condition | Wet Ice |
Starting Material | Plasma, Serum |
Final Product Type | Protein |
Format | Mini Spin Column |
Workflow Step | Protein Depletion |
Quantity | 24 Columns |
Unit Size | 24 columns |
Description | HSA/Immunoglobulin Depletion Mini Spin Columns |
For Use With (Equipment) | Mass Spectormeter |
Product Type | Column |
Product Line | High Select™ |
Label or Dye | Unlabeled |
Shipping Condition | Wet Ice |
Starting Material | Plasma, Serum |
Final Product Type | Protein |
Format | Mini Spin Column |
Workflow Step | Protein Depletion |
Quantity | 6 Columns |
Unit Size | 6 columns |
Description | HSA/Immunoglobulin Depletion Mini Spin Columns |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Column |
Product Line | High Select™ |
Label or Dye | Unlabeled |
Shipping Condition | Wet Ice |
Starting Material | Plasma, Serum |
Final Product Type | Protein |
Format | Mini Spin Column |
Workflow Step | Protein Depletion |
Quantity | 24 Columns |
Unit Size | 24 columns |
Description | HSA/Immunoglobulin Depletion Midi Spin Columns |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Column |
Product Line | High Select™ |
Label or Dye | Unlabeled |
Shipping Condition | Wet Ice |
Starting Material | Plasma, Serum |
Final Product Type | Protein |
Format | Mini Spin Column |
Workflow Step | Protein Depletion |
Quantity | 10 Columns |
Unit Size | 10 columns |
Description | HSA/Immunoglobulin Depletion Resin |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Resin |
Product Line | High Select™ |
Label or Dye | Unlabeled |
Shipping Condition | Wet Ice |
Starting Material | Plasma, Serum |
Final Product Type | Protein |
Workflow Step | Protein Depletion |
Quantity | 50 mL |
Unit Size | 50 mL |
Description | Top14 Abundant Protein Depletion Midi Spin Columns |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Column |
Product Line | High Select™ |
Label or Dye | Unlabeled |
Shipping Condition | Wet Ice |
Starting Material | Plasma, Serum |
Final Product Type | Protein |
Format | Mini Spin Column |
Workflow Step | Protein Depletion |
Quantity | 10 Columns |
Unit Size | 10 columns |
Description | Top14 Abundant Protein Depletion Resin |
For Use With (Equipment) | Mass Spectormeter |
Product Type | Resin |
Product Line | High Select™ |
Label or Dye | Unlabeled |
Shipping Condition | Wet Ice |
Starting Material | Plasma, Serum |
Final Product Type | Protein |
Workflow Step | Protein Depletion |
Quantity | 50 mL |
Form | Liquid |
Unit Size | 50 mL |
Description | Top14 Abundant Protein Depletion Mini Spin Columns |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Column |
Product Line | High Select™ |
Label or Dye | Unlabeled |
Shipping Condition | Wet Ice |
Starting Material | Plasma, Serum |
Final Product Type | Protein |
Format | Mini Spin Column |
Workflow Step | Protein Depletion |
Quantity | 6 Columns |
Unit Size | 6 columns |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A36370 | 24 columns | Top14 Abundant Protein Depletion Mini Spin Columns | 907.65 Online price 974.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A36365 | 6 columns | HSA/Immunoglobulin Depletion Mini Spin Columns | 234.65 Online price 252.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A36366 | 24 columns | HSA/Immunoglobulin Depletion Mini Spin Columns | 687.65 Online price 714.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A36367 | 10 columns | HSA/Immunoglobulin Depletion Midi Spin Columns | 874.65 Online price 936.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A36368 | 50 mL | HSA/Immunoglobulin Depletion Resin | 2,718.65 Online price 2,910.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A36371 | 10 columns | Top14 Abundant Protein Depletion Midi Spin Columns | 1,119.65 Online price 1,204.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A36372 | 50 mL | Top14 Abundant Protein Depletion Resin | 3,231.65 Online price 3,475.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A36369 | 6 columns | Top14 Abundant Protein Depletion Mini Spin Columns | 303.65 Online price 326.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|